Skip to Content
Merck
All Photos(1)

Key Documents

F9005

Supelco

Flufenamic acid

analytical standard, for drug analysis

Synonym(s):

N-(3-[Trifluoromethyl]phenyl)anthranilic acid, N-(α,α,α-Trifluoro-m-tolyl)anthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CF3C6H4NH)C6H4CO2H
CAS Number:
Molecular Weight:
281.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-135 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F

InChI

1S/C14H10F3NO2/c15-14(16,17)9-4-3-5-10(8-9)18-12-7-2-1-6-11(12)13(19)20/h1-8,18H,(H,19,20)

InChI key

LPEPZBJOKDYZAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flufenamic acid, a non-steroidal anti-inflammatory drug, is an effective analgesic agent.

Application

Flufenamic acid has been used as a standard in studying the two well defined, polymorphs of flufenamic acid like form I and form III, using solid-state density functional theory and terahertz spectroscopy.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia C Schwarz et al.
International journal of pharmaceutics, 437(1-2), 83-88 (2012-08-21)
Nanocarriers are highly interesting delivery systems for the dermal application of drugs. Based on a eudermic alkylpolyglycosid nanoemulsions, solid lipid nanoparticles (SLN) and nano-structured lipid carriers (NLC) were prepared by ultrasonic dispersion. The ultrasound preparation technique turned out to be
Minsoo Kim et al.
Pharmaceutics, 11(3) (2019-03-22)
Concomitant use of rivaroxaban with non-dihydropyridine calcium channel blockers (non-DHPs) might lead to an increase of systemic rivaroxaban exposure and anticoagulant effects in relation to the inhibition of metabolic enzymes and/or transporters by non-DHPs. This study was designed to evaluate
Julia C Schwarz et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 81(3), 557-562 (2012-05-09)
Microemulsions are thermodynamically stable, colloidal drug delivery systems. This study presents the first substantiated comparison of natural, skin-compatible and biodegradable surfactants in terms of their suitability to form isotropic microemulsions and their skin interaction. Pseudoternery phase diagrams were constructed for
Antonella Di Pizio et al.
Cellular and molecular life sciences : CMLS, 77(3), 531-542 (2019-06-27)
Human bitter taste receptors (TAS2Rs) are a subfamily of 25 G protein-coupled receptors that mediate bitter taste perception. TAS2R14 is the most broadly tuned bitter taste receptor, recognizing a range of chemically diverse agonists with micromolar-range potency. The receptor is
Conformational origins of polymorphism in two forms of flufenamic acid
Delaney P S, et al.
Journal of Molecular Structure, 1078, 83-89 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service