Skip to Content
Merck
All Photos(1)

Key Documents

D4011

Sigma-Aldrich

N,N-Dimethyl-1-naphthylamine

≥98.0% purity (GC), liquid

Synonym(s):

1-Dimethylaminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7N(CH3)2
CAS Number:
Molecular Weight:
171.24
Beilstein:
1424075
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

N,N-Dimethyl-1-naphthylamine, ≥98.0% (GC)

Quality Level

Assay

≥98.0% (GC)

form

liquid

technique(s)

titration: suitable

color

faint yellow to dark yellow

refractive index

n20/D 1.622 (lit.)

bp

139-140 °C/13 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1cccc2ccccc12

InChI

1S/C12H13N/c1-13(2)12-9-5-7-10-6-3-4-8-11(10)12/h3-9H,1-2H3

InChI key

AJUXDFHPVZQOGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye.

Application

N,N-Dimethyl-1-naphthylamine (1-Dimethylaminonaphthalene) is used to study nitrate reduction by organisms. Reduction of nitrate results in the formation of nitrite, which reacts with sulfanilic acid and 1-dimethylaminonaphthalene to form a red diazo dye.

Biochem/physiol Actions

N,N-Dimethyl-1-naphthylamine (1-Dimethylaminonaphthalene) is used to study nitrate reduction by organisms. Reduction of nitrate results in the formation of nitrite, which reacts with sulfanilic acid and 1-dimethylaminonaphthalene to form a red diazo dye.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Washington JA
Laboratory Procedures in Clinical Microbiology (2012)
L Ingham et al.
Zentralblatt fur Bakteriologie : international journal of medical microbiology, 279(2), 225-230 (1993-06-01)
A rapid swab method using either a dry or moist swab soaked with potassium nitrate/benzalkonium chloride solution, and a method employing a single powdered reagent were assessed as possible alternatives to the conventional test for the detection of bacterial nitrate
[Sensitivity of methods for determining nitrites].
P A Kal'tianis et al.
Laboratornoe delo, (3)(3), 27-28 (1982-01-01)
Chiharu Mizuguchi et al.
Biochimica et biophysica acta, 1841(12), 1716-1724 (2014-10-05)
Human apolipoprotein E (apoE) isoforms exhibit different conformational stabilities and lipid-binding properties that give rise to altered cholesterol metabolism among the isoforms. Using Trp-substituted mutations and site- directed fluorescence labeling, we made a comprehensive comparison of the conformational organization of
Jonas Camillus Jeppesen et al.
Langmuir : the ACS journal of surfaces and colloids, 31(46), 12699-12707 (2015-10-27)
Gel domains in lipid bilayers are structurally more complex than fluid domains. Growth dynamics can lead to noncircular domains with a heterogeneous orientational texture. Most model membrane studies involving gel domain morphology and lateral organization assume the domains to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service