Skip to Content
Merck
All Photos(1)

Key Documents

D0406200

Deptropine citrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dibenzheptropine citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO · C6H8O7
CAS Number:
Molecular Weight:
525.59
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

deptropine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C23H27NO.C6H8O7/c1-24-18-12-13-19(24)15-20(14-18)25-23-21-8-4-2-6-16(21)10-11-17-7-3-5-9-22(17)23;7-3(8)1-6(13,5(11)12)2-4(9)10/h2-9,18-20,23H,10-15H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

CHQGYMXXKZPWOI-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Deptropine citrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DIBENZHEPTROPINE CITRATE ("BRONTINA")--A DOUBLE BLIND TRIAL IN CHRONIC CHEST DISEASE.
M W MCNICOL et al.
British journal of diseases of the chest, 58, 135-140 (1964-07-01)
Uptake of quaternary ammonium compounds into rat intestinal brush border membrane vesicles.
P G Ruifrok
Biochemical pharmacology, 30(19), 2637-2641 (1981-10-01)
[Alarming increase in deptropine poisonings].
R W Griffioen
Nederlands tijdschrift voor geneeskunde, 140(11), 629-630 (1996-03-16)
[Alarming increase in deptropine poisonings].
S M Dreijer-van der Glas
Nederlands tijdschrift voor geneeskunde, 140(11), 628-629 (1996-03-16)
[The treatment of asthma in childhood].
F G Versteegh
Nederlands tijdschrift voor geneeskunde, 132(2), 85-85 (1988-01-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service