Skip to Content
Merck

Skip To

C8606

Dibasic Calcium Phosphate

meets USP testing specifications

Synonym(s):

Calcium phosphate dibasic, Calcium hydrogen phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CaHPO4
CAS Number:
Molecular Weight:
136.06
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
231-826-1
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

USP/NF, meets USP testing specifications

assay

97.5-102.5% anhydrous basis

form

powder

application(s)

pharmaceutical (small molecule)

SMILES string

[Ca++].OP([O-])([O-])=O

InChI

1S/Ca.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+2;/p-2

InChI key

FUFJGUQYACFECW-UHFFFAOYSA-L


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7263307653C3161
assay

97.5-102.5% anhydrous basis

assay

98.0-105.0%

assay

98%

assay

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

agriculture

form

powder

form

powder

form

powder

form

powder, solid

agency

USP/NF, meets USP testing specifications

agency

-

agency

-

agency

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters