Skip to Content
Merck
All Photos(1)

Key Documents

C1510000

Chlorhexidine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1′-Hexamethylenebis[5-(4-chlorophenyl)biguanide]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)3NHC(=NH)NHC(=NH)NHC6H4Cl]2
CAS Number:
Molecular Weight:
505.45
Beilstein:
2826432
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

chlorhexidine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

134-136 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

InChI

1S/C22H30Cl2N10/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34)

InChI key

GHXZTYHSJHQHIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorhexidine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Price et al.
BMJ (Clinical research ed.), 348, g2197-g2197 (2014-04-02)
To determine the effect on mortality of selective digestive decontamination, selective oropharyngeal decontamination, and topical oropharyngeal chlorhexidine in adult patients in general intensive care units and to compare these interventions with each other in a network meta-analysis. Systematic review, conventional
Matthias Maiwald et al.
PloS one, 7(9), e44277-e44277 (2012-09-18)
Skin antisepsis is a simple and effective measure to prevent infections. The efficacy of chlorhexidine is actively discussed in the literature on skin antisepsis. However, study outcomes due to chlorhexidine-alcohol combinations are often attributed to chlorhexidine alone. Thus, we sought
S Karki et al.
The Journal of hospital infection, 82(2), 71-84 (2012-08-15)
The topical use of chlorhexidine gluconate (CHG) is intended to reduce bacterial density on patients' skin. To assess the impact of body bath or skin cleansing with CHG-impregnated or CHG-saturated washcloths in preventing healthcare-associated infections and colonization. This systematic review
Carolyne Horner et al.
The Journal of antimicrobial chemotherapy, 67(11), 2547-2559 (2012-07-27)
Antiseptic agents are increasingly used for hand hygiene and skin decolonization as key tools for the prevention of healthcare-associated infections. Chlorhexidine, a divalent, cationic biguanide, has a broad spectrum of activity and is one of the most frequently used topical
Steven P Engebretson et al.
JAMA, 310(23), 2523-2532 (2013-12-19)
Chronic periodontitis, a destructive inflammatory disorder of the supporting structures of the teeth, is prevalent in patients with diabetes. Limited evidence suggests that periodontal therapy may improve glycemic control. To determine if nonsurgical periodontal treatment reduces levels of glycated hemoglobin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service