Skip to Content
Merck
All Photos(1)

Key Documents

C0465000

Carbimazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,3-Dihydro-3-methyl-2-thioxo-1H-imidazole-1-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2S
CAS Number:
Molecular Weight:
186.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carbimazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C7H10N2O2S/c1-3-11-7(10)9-5-4-8(2)6(9)12/h4-5H,3H2,1-2H3

InChI key

CFOYWRHIYXMDOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbimazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah M A Caney
Journal of feline medicine and surgery, 15(6), 494-502 (2013-04-18)
Hyperthyroidism is the most common feline endocrinopathy. Treatment options comprise anti-thyroid medication, iodine-restricted diet, surgical thyroidectomy and radioiodine. One hundred and eleven owners of hyperthyroid cats completed a detailed survey asking about their experiences and views on the management of
Dinesh Kumar Dhanwal et al.
The Journal of the Association of Physicians of India, 59, 561-562 (2012-02-18)
Hyperthyroidism is associated with bone loss, which is reversible after treatment. The extent of reversibility of loss of bone mass density (BMD) in hyperthyroid patients after treatment especially at forearm is not clear. Therefore, the present study was conducted to
Which antithyroid drug regimen in paediatric Graves' disease?
Daniel Schenk et al.
Clinical endocrinology, 77(6), 806-807 (2012-07-24)
Dhanya Muralidharan et al.
Oral surgery, oral medicine, oral pathology and oral radiology, 115(5), 617-623 (2013-03-05)
Animal studies have shown thyroid dysfunction affects salivary gland functioning, however conclusive human studies are lacking. We sought to assess the qualitative and quantitative changes in saliva among subjects with thyroid dysfunction prior to and following treatment. A longitudinal observational
Nishikant Damle et al.
Journal of pediatric endocrinology & metabolism : JPEM, 24(11-12), 1051-1054 (2012-02-09)
Autonomously functioning thyroid nodules (AFTN) are extremely rare in children. Malignancy may be rarely found in hyperfunctioning 'hot' nodules in adults. However, there are limited reports of AFTNs in young children presenting as or developing malignancy in future. We report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service