Skip to Content
Merck
All Photos(1)

Key Documents

BCR134

Benzo[c]phenanthrene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909296
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3ccc4ccccc4c23

InChI

1S/C18H12/c1-3-7-16-13(5-1)9-11-15-12-10-14-6-2-4-8-17(14)18(15)16/h1-12H

InChI key

TUAHORSUHVUKBD-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR134

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol D Swartz et al.
Mutation research, 661(1-2), 47-56 (2008-12-02)
Sulfur-containing polycyclic aromatic hydrocarbons (thia-PAHs or thiaarenes) are common constituents of air pollution and cigarette smoke, but only a few have been studied for health effects. We evaluated the mutagenicity in Salmonella TA98, TA100, and TA104 of two sulfur-containing derivatives
R B Roth et al.
Biochemistry, 40(17), 5200-5207 (2001-04-25)
Benzo[c]phenanthrene diol epoxide (B[c]PhDE), the ultimate carcinogenic metabolite of the environmental pollutant benzo[c]phenanthrene, reacts with DNA primarily at the exocyclic amino groups of purines, forming B[c]PhDE-DNA adducts that differ in their stereochemical configurations and their effect on biological processes such
V P Gupta et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(1), 82-101 (2008-10-22)
A systematic study has been conducted on the conformation, electronic structure and vibrational spectra of benzo[c]phenanthrene and some of its partially reduced derivatives by experimental infrared spectroscopic and quantum chemical techniques. Electrostatic potential surfaces have been mapped over the electron
Cédric Brulé et al.
The Journal of organic chemistry, 72(9), 3232-3241 (2007-03-31)
A series of novel carbocations were generated by low-temperature protonation of substituted benzo[c]phenanthrenes, B[c]Phs, and their charge delocalization pathways were elucidated by NMR on the basis of the magnitude of Deltadelta13C values. It has been shown that the protonation regioselectivity
Min Wu et al.
Frontiers in bioscience : a journal and virtual library, 9, 2807-2818 (2004-09-09)
Remarkably different conformations can result when DNA binds with stereoisomeric compounds containing differing absolute configurations of substituents about chiral carbon atoms. Furthermore, the biochemical functions of covalent adducts with DNA are strongly affected by the stereochemistry of the ligands. Such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service