Skip to Content
Merck
All Photos(6)

Documents

B2883

Sigma-Aldrich

bisBenzimide H 33258

≥98% (HPLC and TLC)

Synonym(s):

2′-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi(1H-benzimidazole) trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazoyl]-6-(1-methyl-4-piperazyl)benzimidazole trihydrochloride, BBIH, BXI-72, HOE 33258, Hoechst 33258, NSC334072

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N6O · 3HCl
CAS Number:
Molecular Weight:
533.88
Beilstein:
4088183
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥98% (HPLC and TLC)

form

powder

mp

280 °C

solubility

H2O: 10 mg/mL
water and ethanol: 10 mg/mL
phosphate buffer: precipitates

suitability

passes application test for fluorescence

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].Cl[H].[H]O[H].CN1CCN(CC1)c2ccc3NC(=NCc3c2)c4ccc5NC(=NCc5c4)c6ccc(O)cc6

InChI

1S/C27H28N6O.3ClH.H2O/c1-32-10-12-33(13-11-32)22-5-9-25-21(15-22)17-29-27(31-25)19-4-8-24-20(14-19)16-28-26(30-24)18-2-6-23(34)7-3-18;;;;/h2-9,14-15,34H,10-13,16-17H2,1H3,(H,28,30)(H,29,31);3*1H;1H2

InChI key

OWRSPPSBNWJJAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

bisBenzimide H 33258 is useful for staining DNA, chromosomes and nuclei. bisBenzimide H 33258 may be used for fluorescence microscopy or flow cytometry.
Excitation max. = 346 nm
Emission max. = 460 nm
bisBenzimide H 33258 has been used for nuclear staining in cells.

Biochem/physiol Actions

bisBenzimide H 33258 is a potent and selective Bcl-XL inhibitor. bisBenzimide H 33258 is a membrane-permeable, fluorescent DNA stain with low cytotoxicity that intercalate in A-T regions of DNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spontaneous and continuous cyclooxygenase-2-dependent prostaglandin E2 production by stromal cells in the murine small intestine lamina propria: directing the tone of the intestinal immune response.
Newberry RD, et al.
Journal of Immunology, 166, 4465-4472 (2001)
Susanna Fiorelli et al.
Cells, 8(4) (2019-04-19)
Reactive oxygen species (ROS) induce nuclear factor erythroid 2-related factor 2 (Nrf2) activation as an adaptive defense mechanism, determining the synthesis of antioxidant molecules, including heme-oxygenase-1 (HO-1). HO-1 protects cells against oxidative injury, degrading free heme and inhibiting ROS production.
Florian Ehrlich et al.
Scientific reports, 9(1), 10924-10924 (2019-07-31)
Keratins are the main cytoskeletal proteins of epithelial cells and changes in the expression of keratins have contributed to the evolutionary adaptation of epithelia to different environments. Keratin K24 was proposed to be a differentiation marker of epidermal keratinocytes but
Zev A Binder et al.
Cancer cell, 34(1), 163-177 (2018-07-11)
We explored the clinical and pathological impact of epidermal growth factor receptor (EGFR) extracellular domain missense mutations. Retrospective assessment of 260 de novo glioblastoma patients revealed a significant reduction in overall survival of patients having tumors with EGFR mutations at
Conversion of human umbilical cord mesenchymal stem cells in Wharton's jelly to dopaminergic neurons in vitro: potential therapeutic application for Parkinsonism.
Fu YS, et al.
Stem Cells, 24, 115-124 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service