Skip to Content
Merck
All Photos(2)

Key Documents

A7409

Sigma-Aldrich

Bovine Serum Albumin solution

35% in 0.85% sodium chloride, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

description

contains no preservative

sterility

aseptically filled

form

solution

purified by

heat shock fractionation

origin

USA origin

concentration

33.0-37.4% protein (biuret)
35% in 0.85% sodium chloride

technique(s)

microbiological culture: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin belongs to the four-member albumin superfamily. It is a major protein in the circulating blood. Albumin has a helical structure with three domains that form a heart-like shape.

Application

Bovine Serum Albumin solution has been used:
  • to block protein-coated well plates in enzyme-linked immunosorbent assay (ELISA) for immunoreactivity and cross-reactivity studies in the production of monoclonal antibodies (mAbs)
  • to prepare primary antibody solution for peroxidases staining
  • as a supplement for in vitro cultivation of Borrelia strains

Biochem/physiol Actions

Bovine serum albumin (BSA) serves as a protein supplement for cell culture media. BSA acts as a transport carrier for drugs, and it is commonly used to study the protein-drug interactions. Furthermore, it is known for its ability to interact with several aromatic and hydrophobic molecules of the bloodstream.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatai S Oladunni et al.
Journal of virological methods, 290, 114084-114084 (2021-01-30)
The use of monoclonal neutralizing antibodies (mNAbs) is being actively pursued as a viable intervention for the treatment of Severe Acute Respiratory Syndrome CoV-2 (SARS-CoV-2) infection and associated coronavirus disease 2019 (COVID-19). While highly potent mNAbs have great therapeutic potential
Bovine serum albumin
Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and Interactions, 1045 null
Sabrina Hepner et al.
Ticks and tick-borne diseases, 10(5), 1157-1161 (2019-06-27)
Borrelia turcica is a reptile-associated Borrelia species that is vectored by the hard tick Hyalomma aegyptium. Tortoises of the genus Testudo represent the principal host of adult H. aegyptium, while immature stages are less host-specific and can be found on
Jennifer Zechel et al.
Methods in molecular biology (Clifton, N.J.), 399, 79-98 (2008-03-04)
Mitochondrial dysfunction has been increasingly shown as a critical process that makes certain areas of the brain more susceptible not only to neurological disease but also to aging. Quantitative histochemistry is a series of procedures for measuring select metabolites in
The NMR tube bioreactor
Methods in Enzymology, 71-101 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service