A5348
N-(5-Aminopentyl)biotinamide trifluoroacetate salt
≥98% (TLC), solid
Synonym(s):
Biotin–cadaverine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H29F3N4O4S
Molecular Weight:
442.50
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
solid
mol wt
442.5
color
white to off-white
solubility
DMSO or DMF: soluble
application(s)
hematology
histology
storage temp.
room temp
SMILES string
OC(=O)C(F)(F)F.NCCCCCNC(=O)CCCC[C@@H]1SCC2NC(=O)NC12
InChI
1S/C15H28N4O2S.C2HF3O2/c16-8-4-1-5-9-17-13(20)7-3-2-6-12-14-11(10-22-12)18-15(21)19-14;3-2(4,5)1(6)7/h11-12,14H,1-10,16H2,(H,17,20)(H2,18,19,21);(H,6,7)/t11-,12-,14-;/m0./s1
InChI key
QWKKJNXOPDNIEU-RGESYUBESA-N
Application
Biotinylation reagent
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service