Skip to Content
Merck
All Photos(1)

Key Documents

97622

Supelco

2-Mercaptoethanol

for HPLC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein:
773648
EC Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.05

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

quality

LiChropur

expl. lim.

18 %

concentration

14.3 M (pure liquid)

technique(s)

HPLC: suitable

refractive index

n20/D 1.500 (lit.)
n20/D 1.500-1.502

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Mercaptoethanol is a thiol compound showing growth promoting properties on mouse lymphoma L1210 cells in vitro. It also enhances viability, blast transformation and antibody formation in lymphocyte cultures.

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.
suitable for derivatisation of Amine groups

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of growth stimulation of L1210 cells by 2-mercaptoethanol in vitro. Role of the mixed disulfide of 2-mercaptoethanol and cysteine
Ishii T, et al.
The Journal of Biological Chemistry, 256(23), 12387-12392 (1981)
Leon Tribolet et al.
The Journal of infectious diseases, 211(3), 416-425 (2014-08-21)
Na-ASP-2 is an efficacious hookworm vaccine antigen. However, despite elucidation of its crystal structure and studies addressing its immunobiology, the function of Na-ASP-2 has remained elusive. We probed a 9000-protein human proteome microarray with Na-ASP-2 and showed binding to CD79A
Katharina Rothe et al.
Blood, 123(23), 3622-3634 (2014-04-24)
Previous studies demonstrated that imatinib mesylate (IM) induces autophagy in chronic myeloid leukemia (CML) and that this process is critical to cell survival upon therapy. However, it is not known if the autophagic process differs at basal levels between CML
Emily Cunningham Williams et al.
Human molecular genetics, 23(11), 2968-2980 (2014-01-15)
The disease mechanism of Rett syndrome (RTT) is not well understood. Studies in RTT mouse models have suggested a non-cell-autonomous role for astrocytes in RTT pathogenesis. However, it is not clear whether this is also true for human RTT astrocytes.
Nobuko Akiyama et al.
The Journal of experimental medicine, 211(12), 2425-2438 (2014-11-12)
Medullary thymic epithelial cells (mTECs) expressing the autoimmune regulator AIRE and various tissue-specific antigens (TSAs) are critical for preventing the onset of autoimmunity and may attenuate tumor immunity. However, molecular mechanisms controlling mTEC development remain elusive. Here, we describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service