Skip to Content
Merck
All Photos(1)

Key Documents

92732

Supelco

Trimethylsulfonium hydroxide solution

~0.25 M in methanol, for GC derivatization, LiChropur

Synonym(s):

TMSH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3S(OH)
CAS Number:
Molecular Weight:
94.18
Beilstein:
3910286
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

packaging

pkg of 10 × 1 mL

concentration

~0.25 M in methanol

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

SMILES string

[OH-].C[S+](C)C

InChI

1S/C3H9S.H2O/c1-4(2)3;/h1-3H3;1H2/q+1;/p-1

InChI key

MDTPTXSNPBAUHX-UHFFFAOYSA-M

General description

Trimethylsulfonium hydroxide (TMSH) is a methylating reagent. It can be prepared by dissolving trimethylsulfonium iodide in methanol and water solvent further treating with silver oxide.

Application

TMSH solution has been used as a derivatizing agent in a study to determine the fatty acids in milk samples using thermochemolysis-gas chromatography-mass spectrometry.

Other Notes

Reagent for the methylation of nucleophiles, e.g. carboxylic acids, alcohols, thiols, N-heterocycles; Rapid derivatization of acids for GC by pre-column transesterification of glycerides

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raimund Nagel et al.
Organic & biomolecular chemistry, 15(36), 7566-7571 (2017-09-01)
Bacteria can produce gibberellin plant hormones. While the bacterial biosynthetic pathway is similar to that of plants, the individual enzymes are very distantly related and arose via convergent evolution. The cytochromes P450 (CYPs) that catalyze the multi-step oxidation of the
Lidia Lipińska et al.
Probiotics and antimicrobial proteins, 10(2), 186-200 (2017-11-08)
The antifungal activity of Lactobacillus pentosus ŁOCK 0979 depends both on the culture medium and on the fungal species. In the control medium, the strain exhibited limited antagonistic activity against indicator food-borne molds and yeasts. However, the supplementation of the
Narges Yazdekhasti et al.
Molecular nutrition & food research, 60(2), 358-368 (2015-10-27)
The protective effect of fish consumption on the cardiovascular system has primarily been ascribed to n-3 fatty acids, but data investigating the vascular effects of fish protein consumption are scarce. This study aimed to investigate the vascular impact of fish
K. Yamauchi et al.
The Journal of Organic Chemistry, 44, 638-638 (1979)
W. Butte
Journal of Chromatography A, 261, 142-142 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service