Skip to Content
Merck
All Photos(3)

Key Documents

910090

Sigma-Aldrich

Fluorescent Brightener 28 disodium salt solution

used as a stain and brightening agent

Synonym(s):

Fluostain I, Calcofluor White LRP, Calcofluor White ST, Calcofluor white M2R, Tinopal LPW

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H42N12O10S2Na2
CAS Number:
Molecular Weight:
960.95
MDL number:
UNSPSC Code:
12171501
NACRES:
NA.47

form

liquid

concentration

25% in water

SMILES string

[Na+].[Na+].OCCN(CCO)c1nc(Nc2ccccc2)nc(Nc3ccc(\C=C\c4ccc(Nc5nc(Nc6ccccc6)nc(n5)N(CCO)CCO)cc4S([O-])(=O)=O)c(c3)S([O-])(=O)=O)n1

InChI

1S/C40H44N12O10S2.2Na/c53-21-17-51(18-22-54)39-47-35(41-29-7-3-1-4-8-29)45-37(49-39)43-31-15-13-27(33(25-31)63(57,58)59)11-12-28-14-16-32(26-34(28)64(60,61)62)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52(19-23-55)20-24-56;;/h1-16,25-26,53-56H,17-24H2,(H

InChI key

YJHDFAAFYNRKQE-YHPRVSEPSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent Brightener 28, also known as Calcofluor White M2R or Fluostain I, is a large, anionic, stilbene derivative of 4,4’-diaminostilbene-2,2’-disulphonic acid. It is a non-specific fluorochrome that binds with cellulose and chitin contained in the cell walls of fungi and other organisms and is used as a stain and fluorescent brightening agent. It exhibits an absorption maximum of 350 nm in methanol and fluoresces blue-white when excited in the violet or near ultraviolet. It bears additional triazinyl and phenyl aromatic groups. The dianionic sulfonate is hydrophilic. It is commercially available as a disodium salt.

Application

  • Fluorescent Brightener 28 is used in microscopy as fluorochromes, especially to visualize plant tissues by binding to cellulose.
  • It is used in microbiology for staining pathogenic fungi, fungal cell walls, Candida albicans biofilms, for the automated counting of bacteria and spores, and as a viability stain.
  • It is a rapid method for the detection of many yeasts and pathogenic fungi such as Microsporidium, Acanthamoeba, Pneumocystis, Naegleria, and Balamuthia species.
  • It is used industrially as a fluorescent brightening agent for cellulose and polyamide fabrics, paper, detergents, and soaps.
  • It has been used in the identification and study of the structure and biosynthesis of chitin from the freshwater sponge Spongilla lacustris, and to elucidate the location of chitin in skeletal structures.

Biochem/physiol Actions

The extended molecule of Calcofluor white M2R binds to cellulose but does not enter living cells, and therefore can be used to visualize plant tissues and viable cells. It absorbs ultraviolet radiation to emit blue light and exhibits enhanced fluorescence when bound to chitin.

Features and Benefits

  • Facilitates rapid detection of many pathogenic yeasts and fungi.
  • As a brightening agent, its addition can offset the tendency of white materials to yellow with age.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conn's Biological Stains
Kiernan & Horobin
Conn?s Biological Stains (2002)
Candida albicans Biofilm-Defective Mutants
Mathias L.Richard
Eukaryotic Cell null
Histological and Histochemical Methods: Theory and Practice
J.A. Kiernan
Histological and Histochemical Methods: Theory and Practice (2015)
Hermann Ehrlich et al.
Journal of structural biology, 183(3), 474-483 (2013-07-09)
This work demonstrates that chitin is an important structural component within the skeletal fibers of the freshwater sponge Spongilla lacustris. Using a variety of analytical techniques ((13)C solid state NMR, FT-IR, Raman, NEXAFS, ESI-MS, Morgan-Elson assay and Calcofluor White Staining);
The Multifunctional Beta-Oxidation Enzyme Is Required for Full Symptom Development by the Biotrophic Maize Pathogen Ustilago Maydis
Jana Klose et at.
Eukaryotic Cell null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service