Skip to Content
Merck
All Photos(2)

Key Documents

90957

Supelco

Swertiamarin

analytical standard

Synonym(s):

(4aR,5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-4a-hydroxy1H,3H-pyrano[3,4-c]pyran-1-one, 4,4a,5,6-Tetrahydro-4aα-hydroxy-1-oxo-5β-vinyl-1H,3H-pyrano[3,4-c]pyran-6-yl β-D-glucopyranoside, Swertiamarine, Swertiamaroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
Beilstein:
55278
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

InChI

1S/C16H22O10/c1-2-7-14(24-6-8-13(21)23-4-3-16(7,8)22)26-15-12(20)11(19)10(18)9(5-17)25-15/h2,6-7,9-12,14-15,17-20,22H,1,3-5H2/t7-,9+,10+,11-,12+,14-,15-,16+/m0/s1

InChI key

HEYZWPRKKUGDCR-QBXMEVCASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

M B Patel et al.
Pharmaceutical biology, 49(4), 383-391 (2011-03-12)
Enicostemma hyssopifolium Verdoon (Gentianaceae) has been documented for various therapeutic effects in traditional systems of medicine; the hypoglycemic and hypolipidemic activities are also well reported. Bioactivity guided fractionation of methanol extract of E. hyssopifolium to test the hypothesis that E.
V Jaishree et al.
Journal of ethnopharmacology, 130(1), 103-106 (2010-04-28)
The whole plant of Enicostemma axillare Raynal (Family: Gentianaceae) is used in variety of diseases in traditional Indian system of medicine including hepatic ailments. Swertiamarin isolated from Enicostemma axillare Raynal was evaluated for antioxidant and hepatoprotective activity. Swertiamarin was isolated
Junfeng Wang et al.
Bioscience, biotechnology, and biochemistry, 74(8), 1583-1590 (2010-08-12)
Swertia mussotii Franch has anti-hepatitis activity and contains a high level of iridoid monoterpenoids. The cytochrome P450 monooxygenase (CYP) geraniol 10-hydroxylase (G10H) is thought to play an important role in iridoid monoterpenoid and indole alkaloid biosynthesis. Here we report the
Yoshiyuki Kimura et al.
Fitoterapia, 82(6), 827-833 (2011-05-17)
The Swertia japonica is used clinically as a remedy for gastrointestinal symptoms in Japan. We examined the effects of a S. japonica and swertiamarin on gastric emptying and gastrointestinal motility in atropine-, dopamine-, and 5-hydroxytryptamine (5-HT)-treated mice. All three preparations
H-L Li et al.
Arzneimittel-Forschung, 62(3), 138-144 (2012-01-27)
An LC-MS/MS method was developed for the quantification of swertiamarin (CAS 17388-39-5) in rat plasma and tissues using gentiopicroside as the internal standard (IS). Swertiamarin and an IS were extracted from plasma and tissues by a simple solid-phase extraction (SPE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service