Skip to Content
Merck
All Photos(5)

Key Documents

902160

Sigma-Aldrich

Phosphorus pentoxide

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

product line

ReagentPlus®
Redi-Dri

Assay

99%

form

powder

quality

free-flowing

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

cation traces

As: ≤100 mg/kg
Fe: ≤100 mg/kg
heavy metals (as Pb): ≤0.02%

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phosphorus pentoxide is a deliquescent compound prepared by reacting phosphorus with air. It is a commonly used as dehydrating and condensing agent in organic synthesis.[1]

Application

Dehydrating agent used for halogenation with tetrabutylammonium halides.

Process for Producing Halogenated Heteroaryl Compounds
Phosphorus pentachloride has been used for the vaccum distillation of 1-methyl-2-pyrrolidinone (NMP), which was employed as mobile phase in a chromatographic analysis.[2]

It may be used as one of the reaction components in the synthesis of dichlorine heptoxide (Cl2O7)[3] and transition-metal phosphides (Ni2P, Co2P and MoP).[4] P2O5 supported on alumina can be used for the solvent-free and microwave-assisted preparation of 1, 5-benzodiazepine analogs.[5] P2O5/KX (X = Br, I) reagent system may be used for the transformation of alcohols into the corresponding alkyl iodides and bromides.[6]
Phosphorus pentoxide/methanesulfonic acid (PPMA) may be used as a condensing agent and solvent for the synthesis of:
  • High molecular weight poly(benzoxazole)s via direct polycondensation of aromatic dicarboxylic acids containing phenyl ether structure with 3,3′-dihydroxybenzidine dihydrochloride.[7]
  • Aromatic poly(phenylene ether ether ketone)s via direct self-polycondensation of 4-(4′-phenoxyphenoxy)benzoic acids.[8]
  • 1,3-1H-dibenzimidazole-benzene by the reaction of isophthalic acid with 1,2-diaminobenzene.[9]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of poly (benzoxazole) s by direct polycondensation of dicarboxylic acids with 3, 3??dihydroxybenzidine dihydrochloride using phosphorus pentoxide/methanesulfonic acid as condensing agent and solvent.
Ueda M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 24(5), 1019-1026 (1986)
Synthesis of aromatic poly (ether ketone) s in phosphorus pentoxide/methanesulfonic acid.
Ueda M and Oda M.
Polymer Journal, 21(9), 673-679 (1989)
Kinetics of melting and crystallization of phosphorus pentoxide.
Cormia RL, et al.
Journal of Applied Physics, 34(8), 2239-2244 (1963)
An efficient and selective method for the iodination and bromination of alcohols under mild conditions.
Khazdooz L, et al.
Tetrahedron Letters, 57(2), 168-171 (2016)
Eagleson M.
Concise Encyclopedia Chemistry, 617-617 (1994)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service