Skip to Content
Merck
All Photos(1)

Key Documents

89131

Supelco

Fragrance allergen mix A1

24 components in tert-Butyl methyl ether, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

quality

24 components in tert-Butyl methyl ether

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
in compliance with IFRA method

Application

Application Note: "Allergenic Fragrance Testing – New Certified Reference Materials and GC-FID/GC-MS Application" article from AnalytixReporter issue 3

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    α-Amylcinnamyl alcohol
    trans-Anethole
    4-Methoxybenzyl alcohol
    Benzyl alcohol
    β-Caryophyllene
    Cinnamyl alcohol
    Citronellol
    Ebanol total
    Eugenol
    trans,trans-Farnesol
    Geraniol
    Isoeugenol (E+Z)
    (R)-(+)-Limonene
    Linalool
    Menthol
    (+)-α-Pinene
    (−)-β-Pinene
    β-Santalol
    Sclareol
    α-Terpinene
    Terpineol total
    tert-Butyl methyl ether
    β,β,3-Trimethylbenzenepropanol
    See All (23)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MoCRA brings the most significant changes to US cosmetic regulations in 80 years, requiring companies to monitor FDA updates and adopt new compliance policies.

MoCRA brings the most significant changes to US cosmetic regulations in 80 years, requiring companies to monitor FDA updates and adopt new compliance policies.

MoCRA brings the most significant changes to US cosmetic regulations in 80 years, requiring companies to monitor FDA updates and adopt new compliance policies.

MoCRA brings the most significant changes to US cosmetic regulations in 80 years, requiring companies to monitor FDA updates and adopt new compliance policies.

Protocols

IFRA method introduces new CRMs for allergenic fragrances, analyzed by GC with ionic liquid capillary column for selectivity.

IFRA method introduces new CRMs for allergenic fragrances, analyzed by GC with ionic liquid capillary column for selectivity.

IFRA method introduces new CRMs for allergenic fragrances, analyzed by GC with ionic liquid capillary column for selectivity.

IFRA method introduces new CRMs for allergenic fragrances, analyzed by GC with ionic liquid capillary column for selectivity.

Questions

  1. Bonjour, Mise en place d'un protocole d'analyse des allergènes par CG au sein de mon entreprise, mais nous avons du mal à comprendre pourquoi vous proposez 2 méthodes d'analyses différentes sur vos mix avec 2 colonnes différentes la 29832-U et la 24181

    1 answer
    1. See the link below to review the method recommended for 89131, Fragrance allergen mix A1.
      https://www.sigmaaldrich.com/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/allergenic-fragrance-testing

      Note that the recommended column is 29832-U, SLB®-IL60i 30 m x 0.25 mm x 0.2 μm. The usage of column 24181, SPB®-50 Capillary GC Column, cannot be located. The information you are requesting may be available, however additional information is required. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information, including the reference to the 24181 column, so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service