Skip to Content
Merck
All Photos(1)

Key Documents

86485

Sigma-Aldrich

Terpinolene

technical, ≥85% (GC)

Synonym(s):

4-Isopropylidene-1-methylcyclohexene, p-Menth-1,4(8)-diene, p-Meth-1-en-8-yl-formate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1851203
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~4.7 (vs air)

Quality Level

vapor pressure

~0.5 mmHg ( 20 °C)

grade

technical

Assay

≥85% (GC)

form

liquid

refractive index

n20/D 1.489 (lit.)
n20/D 1.491

bp

184-185 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C1/CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4H,5-7H2,1-3H3

InChI key

MOYAFQVGZZPNRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terpinolene is a biogenically emitted monoterpene. Kinetics of gas phase reaction of OH and NO3 radicals with terpinolene has been investigated. Biotransformation of terpinolene byAspergillus niger and several related species has been reported. Epoxidation of terpinolene in anhydrous conditions using alumina as a heterogeneous catalyst and hydrogen peroxide as the oxidant agent has been studied. It is an major component of essential oil extracted from the aerial parts of Clausena indica.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - Seta closed cup

Flash Point(C)

61 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tran Huy Thaia et al.
Natural product communications, 9(10), 1531-1534 (2014-12-20)
The chemical composition of Vietnamese oil samples of the aerial parts of Clausena indica (Dalz.) Oliver and C. anisum-olens (Blanco) Merryll have been investigated using a combination of chromatographic and spectroscopic techniques. C. indica essential oil contained mainly terpinolene (53.9
Alumina as heterogeneous catalyst for the regioselective epoxidation of terpenic diolefins with hydrogen peroxide.
Uguina MA, et al.
J. Mol. Catal. A: Chem., 256(1), 208-215 (2006)
Kinetics of the gas-phase reactions of hydroxyl and nitrogen oxide (NO3) radicals with 2-carene, 1, 8-cineole, p-cymene, and terpinolene.
Corchnoy SB and Atkinson R.
Environmental Science & Technology Letters, 24(10), 1497-1502 (1990)
Biotransformation of monoterpenoids,(-)-and menthols, terpinolene and carvotanacetone by Aspergillus species.
Asakawa Y, et al.
Phytochemistry, 30(12), 3981-3987 (1991)
Yan Ma et al.
Physical chemistry chemical physics : PCCP, 11(21), 4198-4209 (2009-05-22)
Gas-phase ozonolysis of terpinolene was studied in static chamber experiments using gas chromatography coupled to mass spectrometric and flame ionisation detection to separate and detect products. Two isomers of C(7)-diacids and three isomers of C(7)-aldehydic acids were identified in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service