Skip to Content
Merck
All Photos(1)

Key Documents

85126

Supelco

Silanization solution I

~5% (dimethyldichlorosilane in heptane), Selectophore

Synonym(s):

Dichlorodimethylsilane solution, Dimethyldichlorosilane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6Cl2Si
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

concentration

~5% (dimethyldichlorosilane in heptane)

refractive index

n20/D 1.388

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

dimethyldichlorosilane/heptane 1/30 (v/v)
Ready-to-use solution for silanizing micro-electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Jesús Andrés-Costa et al.
Scientific reports, 7(1), 15777-15777 (2017-11-19)
European legislation focusing on water quality is expected to broaden to encompass several pharmaceuticals as priority hazardous substances. This manuscript aims to challenge current regulatory approaches that do not recognize stereochemistry of chiral pharmaceuticals by testing the hypothesis that environmental
Ion-selective electrode for intracellular potassium measurements.
A Haemmerli et al.
Analytical chemistry, 52(8), 1179-1182 (1980-07-01)
S C Lung et al.
Chemosphere, 41(12), 1865-1871 (2000-11-04)
The effects of coating materials on polychlorinated biphenyl (PCB) adsorption in aqueous solution were assessed in an attempt to minimize PCB sorption loss during sampling processes. A coating material, which enhances PCB adsorption and allows adsorbed PCBs to be readily
Bernard P Binks et al.
Physical chemistry chemical physics : PCCP, 9(48), 6391-6397 (2007-12-07)
We report measured and calculated oil-ionic liquid, water-ionic liquid and oil-water contact angles on silica surfaces which have been hydrophobised to different extents by silanisation. Based on the idea that the contact angle formed by a liquid-liquid interface with a
J Sotres et al.
Journal of dental research, 91(10), 973-978 (2012-08-10)
The pellicle serves as a multifunctional protective layer, providing, e.g., lubrication and remineralization and also acting as a diffusion barrier. In addition, since the formation of the pellicle precedes the adhesion of micro-organisms, it is also important as a conditioning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service