Skip to Content
Merck
All Photos(1)

Key Documents

77968

Supelco

(R)-(+)-α-Methylbenzyl isocyanate

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(R)-(+)-1-Phenylethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NCO
CAS Number:
Molecular Weight:
147.17
Beilstein:
3196930
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D +10.5±0.5°, neat

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.513 (lit.)
n20/D 1.513

bp

55-56 °C/2.5 mmHg (lit.)

density

1.045 g/mL at 20 °C (lit.)
1.046 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

C[C@@H](N=C=O)c1ccccc1

InChI

1S/C9H9NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-6,8H,1H3/t8-/m1/s1

InChI key

JJSCUXAFAJEQGB-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-a-Methylbenzyl isocyanate [(R)-(+)-MBIC] is a derivatizing agent.

Application

(R)-(+)-MBIC has been used in derivatization of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol to form 4-(methylnitrosamino)-1-(a-methylbenzylcarbamoyl)-1-(3-pyridyl)butane. It has also been used in isolating hydroxyl acid, converting it to Methyl-4-[(S)-a-methylbenzylcarbamoyl]-4-(3-pyridyl)- butanoate which was further analysed by LC-MS/MS.1 It reacts with hydrate hydrochloride ketoaniline to yield hemiaminal, 3-((1R)-1-Phenylethyl)-(4S)-6-chloro-4-hydroxy-4-(trifluoromethyl)-1,3,4-trihydroquinazolin-2-one.

Other Notes

Reagent used for the determination of the enantiomeric purity of alcohols and amines

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective metabolism of nicotine and tobacco-specific N-nitrosamines to 4-hydroxy-4-(3-pyridyl)butanoic acid in rats.
Trushin N, Hecht SS.
Chemical Research in Toxicology, 12(2), 164-171 (1999)
R. Dumont et al.
The Journal of Organic Chemistry, 51, 2515-2515 (1986)
Determination of metoprolol enantiomers in plasma and urine using (S)-(-)-phenylethyl isocyanate as a chiral reagent.
G Pflugmann et al.
Journal of chromatography, 421(1), 161-164 (1987-10-09)
General scope of 1,4-diastereoselective additions to a 2(3H)-quinazolinone: practical preparation of HIV therapeutics.
Magnus NA
The Journal of Organic Chemistry, 68(3), 754-761 (2003)
K.C. Rice et al.
The Journal of Organic Chemistry, 45, 592-592 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service