Skip to Content
Merck
All Photos(1)

Key Documents

76607

Supelco

Urethane

analytical standard

Synonym(s):

Carbamic acid ethyl ester, Ethyl carbamate, Ethylurethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOC2H5
CAS Number:
Molecular Weight:
89.09
Beilstein:
635810
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.07 (vs air)

vapor pressure

10 mmHg ( 77.8 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

182-184 °C (lit.)

mp

46-49 °C
48-50 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(N)=O

InChI

1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)

InChI key

JOYRKODLDBILNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Urethane is used as an anesthetic in animals, since it shows minimal effects on cardiovascular and respiratory systems and maintenance of spinal reflexes.[1] It can also be used in the synthesis of poly(urethane urea) segmented block, which can find applications as blood sacs in ventricular assist devices and total artificial hearts.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The anesthetic mechanism of urethane: the effects on neurotransmitter-gated ion channels
Hara, et al.
Anesthesia and Analgesia, 94, 313-318 (2002)
New biomedical poly (urethane urea)? layered silicate nanocomposites
Xu R, et al.
Macromolecules, 34, 337-339 (2001)
C A Maggi et al.
Experientia, 42(5), 531-537 (1986-05-15)
The suitability of urethane anesthesia for physiopharmacological experiments in various systems is briefly reviewed. Urethane anesthesia appears to be suitable for various types of studies on respiratory function and on reflex activation of motility of the urinary bladder and some
A A Scopinho et al.
British journal of pharmacology, 155(5), 752-756 (2008-07-22)
Control of food intake is a complex behaviour which involves many interconnected brain structures. The present work assessed if the noradrenergic system in the lateral septum (LS) was involved in the feeding behaviour of rats. In the first protocol, the
Hironori Satoh et al.
Cancer research, 73(13), 4158-4168 (2013-04-24)
Nrf2 (Nfe2l2) governs cellular defenses against oxidative and electrophilic stresses and protects against chemical carcinogenesis. However, many cancers have been found to accumulate NRF2 protein, raising questions of precisely how Nrf2 contributes to carcinogenesis. In this report, we explored such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service