Skip to Content
Merck
All Photos(1)

Documents

76112

Supelco

Deltamethrin-(phenoxy-d5)

PESTANAL®, analytical standard

Synonym(s):

[(S)-Cyano-(3-phenoxy-d5-phenyl)methyl] (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22D5H14Br2NO3
Molecular Weight:
510.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

98-102 °C

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=C([C@@H]1[C@@H](C1(C)C)C=C(Br)Br)O[C@H](C#N)C2=CC=CC(OC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])=C2

InChI

1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1/i3D,4D,5D,8D,9D

InChI key

OWZREIFADZCYQD-LPDJBGJGSA-N

General description

Deltamethrin-(phenoxy-d5) is an isotope-labeled analog of the pyrethroid insecticide deltamethrin, wherein phenoxy protons are substituted by deuterium.

Application

Deltamethrin-(phenoxy-d5) is used as a deuterated internal standard to quantify deltamethrin in goat dairy products, animal fat, and surface water by chromatography techniques.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Deltamethrin Residues in Milk and Cheese of Lactating Goats (Capra hircus)
Marrone R, et al.
Molecules (Basel), 24(3), 517-517 (2019)
Determination of pyrethrin and pyrethroid residues in animal fat using liquid chromatography coupled to tandem mass spectrometry
Moloney M, et al.
Journal of Chromatography. B, Biomedical Applications, 1077(3), 60-70 (2018)
Picogram per liter quantification of pyrethroid and organophosphate insecticides in surface waters: a result of large enrichment with liquid-liquid extraction and gas chromatography coupled to mass spectrometry using atmospheric pressure chemical ionization
Rosch A, et al.
Analytical and Bioanalytical Chemistry, 411(14), 3151-3164 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service