Skip to Content
Merck
All Photos(1)

Key Documents

76048

Supelco

Dimethyl sulfide

analytical standard

Synonym(s):

DMS, Methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
Beilstein:
1696847
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

2.1 (vs air)

vapor pressure

26.24 psi ( 55 °C)
7.79 psi ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

402 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

19.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

38 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: soluble 7.28 g/L at 20 °C

density

0.846 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CSC

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfide is found to be the most abundant volatile thioether[1], which can find applications as a flavor enhancing agent in food.[2] It can undergo oxidation in the process of malting and kilning, to yield dimethyl sulfoxide (DMSO).[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of l-Cysteine and Transition Metal Ions on Dimethyl Sulfide Oxidation
Baldus M, et al.
Journal of Agricultural and Food Chemistry, 65(10), 2180-2188 (2017)
Dimethyl sulfide in the surface ocean and the marine atmosphere: a global view
Andreae.OM and Raemdonck H
Science, 744-747 (1983)
Tae Gwan Kim et al.
Applied microbiology and biotechnology, 97(7), 3171-3181 (2012-05-11)
Methanotrophs must become established and active in a landfill biocover for successful methane oxidation. A lab-scale biocover with a soil mixture was operated for removal of methane and nonmethane volatile organic compounds, such as dimethyl sulfide (DMS), benzene (B), and
K S Carslaw et al.
Nature, 503(7474), 67-71 (2013-11-10)
The effect of anthropogenic aerosols on cloud droplet concentrations and radiative properties is the source of one of the largest uncertainties in the radiative forcing of climate over the industrial period. This uncertainty affects our ability to estimate how sensitive
Sheree Yau et al.
The ISME journal, 7(10), 1944-1961 (2013-04-27)
Organic Lake is a shallow, marine-derived hypersaline lake in the Vestfold Hills, Antarctica that has the highest reported concentration of dimethylsulfide (DMS) in a natural body of water. To determine the composition and functional potential of the microbial community and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service