Skip to Content
Merck
All Photos(2)

Key Documents

75934

Supelco

3β-Taraxerol

analytical standard

Synonym(s):

(3β)-D-Friedoolean-14-en-3-ol, (3β13α)-13-Methyl-27-norolean-14-en-3-ol, Alnulin, Skimmiol, Taraxerol, Tiliadin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)CC[C@]2(C)CC=C3[C@]4(C)CC[C@@]5([H])C(C)(C)[C@@H](O)CC[C@]5(C)[C@@]4([H])CC[C@@]3(C)[C@]2([H])C1

InChI

1S/C30H50O/c1-25(2)17-18-27(5)13-9-21-29(7)14-10-20-26(3,4)24(31)12-16-28(20,6)22(29)11-15-30(21,8)23(27)19-25/h9,20,22-24,31H,10-19H2,1-8H3/t20-,22+,23+,24-,27-,28-,29-,30+/m0/s1

InChI key

GGGUGZHBAOMSFJ-GADYQYKKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ru-Mei Lu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 32(7), 1056-1059 (2009-10-31)
To study the chemical constituents of Uvaria microcarpa. The constituents were repeatedly separated and purified with silica gel column and Sephadex LH-20 column, and identified by physico-chemical properties and spectral methods. Nine compounds were separated and identified as beta-sitosterol palmitate
Jaya Shahi et al.
Revista do Instituto de Medicina Tropical de Sao Paulo, 53(5), 259-263 (2011-10-21)
Channa punctatus was exposed to four different concentrations of Rutin, Taraxerol and Apigenin. Changes in some hematological parameters of Channa punctatus were assessed to determine the influence of these compounds on test fish. Fish were exposed to sublethal concentrations (80%
Yan Shi et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(16), 1994-1996 (2008-12-18)
To study the chemical constituents from the roots of Pterospermum heterophyllum. The chemical constituents of P. heterophyllum were isolated and purified by silica gel and sephadex LH-20 column chromatography. Their structures were identified on the basis of spectroscopic data and
Kadapakkam Nandabalan Sangeetha et al.
Biochimica et biophysica acta, 1800(3), 359-366 (2009-12-23)
The present study focuses on identifying and developing an anti-diabetic molecule from plant sources that would effectively combat insulin resistance through proper channeling of glucose metabolism involving glucose transport and storage. Insulin-stimulated glucose uptake formed the basis for isolation of
Jian Wei et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 30(1), 47-49 (2007-06-02)
7 compounds were isolated from the ethyl-acetate extract of Vaccinium iteophyllum Hance by using repeated silical gel column chromatography. These 7 compounds were identified by means of physico-chemical propertic and spectroscopic analysis as beta-sitosterol (I), ursolic acid (II), taraxerol (III)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service