Skip to Content
Merck
All Photos(1)

Key Documents

75054

Supelco

Valeric acid

analytical standard

Synonym(s):

n-Valeric acid, Pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
969454
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)

Assay

≥99.8% (GC)

autoignition temp.

707 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.408 (lit.)
n20/D 1.408

bp

110-111 °C/10 mmHg (lit.)
185 °C (lit.)

mp

−20-−18 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCC(O)=O

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valeric acid is a renewable chemical that can be obtained via hydrogenation of levulinic acid in the presence of nickel-based catalyst or from lignocellulosic biomass. It can undergo esterification to form valerate esters, which can find applications as blending agents for gasoline and diesel, used in the production of biofuels.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of Valeric Acid Ketonization and Ketenization in Catalytic Pyrolysis on Nanosized SiO2, ?-Al2O3, CeO2/SiO2, Al2O3/SiO2 and TiO2/SiO2
Kulyk K, et al.
ChemPhysChem (2017)
Influence of W on the reduction behaviour and Br?nsted acidity of Ni/TiO 2 catalyst in the hydrogenation of levulinic acid to valeric acid: Pyridine adsorbed DRIFTS study
Kumar.VV, et al.
Applied Catalysis A: General, 531, 169-176 (2017)
M F Böttcher et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 30(11), 1590-1596 (2000-11-09)
The prevalence of allergic diseases has increased particularly over the past 30-40 years. A reduced microbial stimulation during infancy may result in a development of a disturbed balance between Th1- and Th2-like immunity. The gut flora is, quantitatively, the most
D M Saunders et al.
The Australian & New Zealand journal of obstetrics & gynaecology, 18(3), 198-201 (1978-08-01)
The effect of oestradiol valerate on levels of blood cholesterol, triglycerides, free oestradiol and coagulation factors, as well as on urinary oestrogens and free cortisol was measured in 10 oöphorectomised women. Despite the very high urinary oestrogen levles obtained, there
Valeric biofuels: a platform of cellulosic transportation fuels.
Jean-Paul Lange et al.
Angewandte Chemie (International ed. in English), 49(26), 4479-4483 (2010-05-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service