Skip to Content
Merck
All Photos(1)

Key Documents

72706

Sigma-Aldrich

L-Leucine

tested according to Ph. Eur.

Synonym(s):

Leucinum, (S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

Assay

98.5-101.0%

form

powder

color

white

mp

>300 °C (lit.)

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William I A Haynes et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(11), 4804-4814 (2013-03-15)
The identification of a hyperdirect cortico-subthalamic nucleus connection highlighted the important role of the subthalamic nucleus (STN) in regulating behavior. However, this pathway was shown primarily from motor areas. Hyperdirect pathways associated with cognitive and motivational cortical regions are particularly
M Sakamoto et al.
The Journal of clinical endocrinology and metabolism, 56(1), 202-204 (1983-01-01)
Recent studies on the nucleotide sequence of cloned DNA complementary to mRNA for preproenkephalin from bovine adrenal medulla and human pheochromocytoma have revealed that this precursor molecule contains four copies of methionine-enkephalin (Met-Enk) and one copy each of leucine-enkephalin (Leu-Enk)
Qian Wang et al.
Journal of the National Cancer Institute, 105(19), 1463-1473 (2013-09-21)
L-type amino acid transporters (LATs) uptake neutral amino acids including L-leucine into cells, stimulating mammalian target of rapamycin complex 1 signaling and protein synthesis. LAT1 and LAT3 are overexpressed at different stages of prostate cancer, and they are responsible for
Sebastian Kalamajski et al.
Matrix biology : journal of the International Society for Matrix Biology, 29(4), 248-253 (2010-01-19)
Small leucine-rich proteoglycans/proteins (SLRPs) are associated with collagen fibril formation, and therefore important for the proper formation of extracellular matrices. SLRPs are differentially expressed in tissues and during pathological conditions, contributing to the development of connective tissue properties. The binding
T Yoshimasa et al.
The Journal of clinical investigation, 69(3), 643-650 (1982-03-01)
To elucidate the physiological and pathophysiological significance of methionine- and leucine-enkephalin (Met-and Leu-enkephalin, respectively) in human sympathoadrenal system, the contents of these peptides in normal human sympathetic nervous system, adrenal medulla, and pheochromocytomas were determined by specific radioimmunoassays combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service