Skip to Content
Merck
All Photos(2)

Key Documents

72576

Supelco

Myricetin

analytical standard

Synonym(s):

3,3′,4′,5,5′,7-Hexahydroxyflavone, Cannabiscetin, Myricetol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O8
CAS Number:
Molecular Weight:
318.24
Beilstein:
332331
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3cc(O)c(O)c(O)c3

InChI

1S/C15H10O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,16-20,22H

InChI key

IKMDFBPHZNJCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: eucalyptus hypericum primula

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
Saila Vepsäläinen et al.
The Journal of nutritional biochemistry, 24(1), 360-370 (2012-09-22)
A growing body of epidemiological evidence suggests that fruit and vegetable juices containing various phenolic compounds can reduce the risk of Alzheimer's disease (AD). As the altered amyloid precursor protein (APP) processing leading to increased β-amyloid (Aβ) production is a
Neelamegam Kandasamy et al.
Toxicology and applied pharmacology, 279(2), 173-185 (2014-06-14)
Diabetic nephropathy is the kidney disease that occurs as a result of diabetes. The present study was aimed to evaluate the therapeutic potential of myricetin by assaying the activities of key enzymes of carbohydrate metabolism, insulin signaling molecules and renal
Catarina L Silva et al.
Analytica chimica acta, 739, 89-98 (2012-07-24)
A new approach based on microextraction by packed sorbent (MEPS) and reversed-phase high-throughput ultra high pressure liquid chromatography (UHPLC) method that uses a gradient elution and diode array detection to quantitate three biologically active flavonols in wines, myricetin, quercetin, and
Leslie K Williams et al.
Journal of medicinal chemistry, 55(22), 10177-10186 (2012-10-12)
The increasing prevalence of diabetes has accelerated the search for new drugs derived from natural sources. To define the functional features of two such families of compounds, the flavonols and the ethyl caffeates, we have determined the high-resolution structures of

Protocols

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service