Skip to Content
Merck
All Photos(1)

Key Documents

68486

Supelco

Diazinon

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Dimpylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N2O3PS
CAS Number:
Molecular Weight:
304.35
Beilstein:
273790
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

-10 to -25°C

SMILES string

CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)C

InChI

1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3

InChI key

FHIVAFMUCKRCQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

219.9 °F - closed cup

Flash Point(C)

104.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Huen et al.
Environmental research, 117, 8-16 (2012-06-12)
Organophosphate pesticides are widely used and recent studies suggest associations of in utero exposures with adverse birth outcomes and neurodevelopment. Few studies have characterized organophosphate pesticides in human plasma or established how these levels correlate to urinary measurements. We measured
Xiao Zhang et al.
Environmental toxicology and pharmacology, 34(3), 959-968 (2012-09-12)
Pesticide exposure has repeatedly been associated with cancers. However, molecular mechanisms are largely undetermined. In this study, we examined whether exposure to diazinon, a common organophosphate that has been associated with cancers, could induce DNA methylation alterations. We conducted genome-wide
Christina Tsitsimpikou et al.
Toxicology, 307, 109-114 (2012-12-04)
The aim of the present study was to investigate the effects of diazinon and propoxur on liver and kidneys, following long term exposure of rabbits. Ten New Zealand white female rabbits were used. The animals were divided into 5 groups
Dong Wang et al.
Reproductive toxicology (Elmsford, N.Y.), 34(4), 489-497 (2012-09-05)
The potential toxicity resulting from combinatorial effects of organophosphorus and pyrethroid insecticides are not completely known. We evaluated male reproductive toxicity in mice co-exposed to diazinon and cis-permethrin. Nine-week-old male Sv/129 mice were exposed to diazinon (10 μmol/kg/day) or cis-permethrin
Mariusz Cycoń et al.
Journal of environmental management, 117, 7-16 (2013-01-22)
The ability of diazinon-degrading Serratia marcescens to remove organophosphorus pesticides (OPPs), i.e. chlorpyrifos (CP), fenitrothion (FT), and parathion (PT) was studied in a mineral salt medium (MSM) and in three soils of different characteristics. This strain was capable of using

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service