Skip to Content
Merck
All Photos(1)

Documents

63965

Supelco

(−)-β-Elemene

analytical standard

Synonym(s):

β-Elemene, (1S,2S,4R)-(−)-2,4-Diisopropenyl-1-methyl-1-vinylcyclohexane, (1S,2S,4R)-1-Ethenyl-1-methyl-2,4-bis(1-methylethenyl)cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2207781
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@]1(C=C)CC[C@@H](C(C)=C)C[C@H]1C(C)=C

InChI

1S/C15H24/c1-7-15(6)9-8-13(11(2)3)10-14(15)12(4)5/h7,13-14H,1-2,4,8-10H2,3,5-6H3/t13-,14+,15-/m1/s1

InChI key

OPFTUNCRGUEPRZ-QLFBSQMISA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: achillea

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F

Flash Point(C)

98 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huatang Wang et al.
Environmental entomology, 48(1), 245-252 (2018-12-20)
For insects, odorant-binding proteins (OBPs) play an essential role in binding and transporting semiochemicals through the sensillum lymph to olfactory receptor neurons within the antennal sensilla. In the present study, the full-length cDNA encoding a general odorant-binding protein 1 (DcitOBP1
Shuiping Liu et al.
Cell death & disease, 11(11), 969-969 (2020-11-13)
N6-methyladenosine (m6A) modification can alter gene expression by regulating RNA splicing, stability, translocation, and translation. Emerging evidence shows that m6A modification plays an important role in cancer development and progression, including cell proliferation, migration and invasion, cell apoptosis, autophagy, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service