Skip to Content
Merck
All Photos(1)

Key Documents

63954

Supelco

Ethyl acetoacetate

analytical standard

Synonym(s):

Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
Beilstein:
385838
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

4.48 (vs air)

vapor pressure

1 mmHg ( 28.5 °C)

Assay

≥99.0% (GC)

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n/D 1.418-1.420

bp

181 °C (lit.)

mp

−43 °C (lit.)

solubility

water: soluble 130 g/L at 20 °C

density

1.029 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetoacetate is derived from ethanol.[1]

Application

Ethyl acetoacetate may be used:
  • As a diluent for improving cold flow properties of biodiesel from waste cooking oil.[1]
  • As an additive for activating Fe3O4 nanoparticles for efficient catalytic oxidation of aldehydes to carboxylic acids.[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

164.3 °F - closed cup

Flash Point(C)

73.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ethyl acetoacetate: A potential bio-based diluent for improving the cold flow properties of biodiesel from waste cooking oil.
Cao L, et al.
Applied Energy, 114, 18-21 (2014)
Fe3O4 nanoparticles/ethyl acetoacetate system for the efficient catalytic oxidation of aldehydes to carboxylic acids.
Villano R, et al.
Tetrahedron Letters, 55(15), 2442-2445 (2014)
Igor R Costa et al.
Nutrients, 7(1), 1-16 (2014-12-30)
Essential amino acids (EAA) consist of a group of nine amino acids that animals are unable to synthesize via de novo pathways. Recently, it has been found that most metazoans lack the same set of enzymes responsible for the de
P Priscilla Lui
Psychological bulletin, 141(2), 404-446 (2014-12-22)
Among immigrant Asian and Latino groups, the contrast between collectivism in traditional heritage and individualism in the mainstream American cultures presents unique challenges for their family relationships. This systematic review was designed to answer 3 fundamental questions: to what extent
Pernille R Jensen et al.
International journal of cancer, 136(4), E117-E126 (2014-08-27)
An increased prevalence of liver diseases such as hepatitis C and nonalcoholic fatty liver results in an augmented incidence of the most common form of liver cancer, hepatocellular carcinoma (HCC). HCC is most often found in the cirrhotic liver and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service