Skip to Content
Merck
All Photos(1)

Key Documents

63445

Sigma-Aldrich

Almond oil from Prunus dulcis

tested according to Ph. Eur.

Synonym(s):

Amygdalae oleum raffinatum, Almond oil, sweet

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

density

0.91 g/mL at 20 °C

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

P Ferraro et al.
Nature nanotechnology, 5(6), 429-435 (2010-05-11)
Manipulating and dispensing liquids on the micrometre- and nanoscale is important in biotechnology and combinatorial chemistry, and also for patterning inorganic, organic and biological inks. Several methods for dispensing liquids exist, but many require complicated electrodes and high-voltage circuits. Here
Emilia Peleva et al.
The Laryngoscope, 121(12), 2661-2664 (2011-11-24)
Almond oil is frequently prescribed as a ceruminolytic, to soften ear wax or relieve ventilation tube occlusion. Ceruminolytics could lead to ototoxicity in the presence of a tympanic perforation. Reports on the safety of almond oil as a ceruminolytic is
Ossama Kodad et al.
Journal of agricultural and food chemistry, 57(22), 10809-10813 (2009-11-04)
The increasing utilization of self-compatible almond cultivars in solid plantings of a single genotype has raised the question of the effect of the pollen source on the kernel quality of these new autogamous cultivars. Thus, the effect of two different
Aylin Altan et al.
Journal of food science, 76(6), E489-E494 (2012-03-16)
Oil migration from high oil content almond confections into adjacent chocolate causes changes in product quality. The objective of this study was to quantify the oil migration from almond products to dark chocolate. Magnetic resonance imaging (MRI) was used to
Sophie Gallier et al.
Food & function, 3(5), 547-555 (2012-02-23)
An aqueous suspension of almond oil bodies (about 10% lipids) was prepared and subjected to in vitro gastric (with pepsin) and intestinal (with bile salts and pancreatin) digestion, simulating fasting conditions. The physicochemical and structural changes of the almond oil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service