Skip to Content
Merck
All Photos(1)

Key Documents

62155

Supelco

Methyl linolelaidate

analytical standard

Synonym(s):

Linolelaidic acid methyl ester, Methyl trans,trans-9,12-octadecadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H34O2
CAS Number:
Molecular Weight:
294.47
Beilstein:
1727615
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C\C\C=C\CCCCCCCC(=O)OC

InChI

1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7+,11-10+

InChI key

WTTJVINHCBCLGX-ZDVGBALWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasutaka Shimotori et al.
Carbohydrate research, 359, 11-17 (2012-08-29)
Various glycosyl ferulates were efficiently synthesized from 2,4,6-tetra-O-acetyl-α-D-glucopyranosyl bromide (TAGB) with amine by amine-promoted glycosylation without using heavy metal. The resulted acetylated glycosyl ferulates with acetoxyl groups at C-2, C-3 and C-4 were regioselectively deacetylated at C-4 and C-6 positions
Fumie Saito et al.
Bioscience, biotechnology, and biochemistry, 74(1), 168-174 (2010-01-09)
gamma-Tocopherol was reacted with (E)-4-oxo-2-nonenal (ONE) at 37 degrees C in an acidic acetonitrile solution. The reaction products were isolated by reversed-phase high-performance liquid chromatography and their structures were characterized to be 5-substituted gamma-tocopherols: 5-(1-(furan-2-yl)pentyl)-gamma-tocopherol (1), 3-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2yl)propanal (2), and 1-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2-yl)-4-(furan-2-yl)octan-3-one
Philip T Lynett et al.
Organic & biomolecular chemistry, 9(9), 3320-3330 (2011-03-30)
It has long been recognized that garlic and petiveria, two plants of the Allium genus--which also includes onions, leeks and shallots--possess great medicinal value. In recent times, the biological activities of extracts of these plants have been ascribed to the
Guo-Xiang Li et al.
European journal of medicinal chemistry, 45(5), 1821-1827 (2010-02-04)
5,7-, 5,8-, 6,8-, 7,8-dichloro-4-quinolinol-3-carboxylic acid (5,7-, 5,8-, 6,8-, 7,8-DCQA) together with 7-chloro-4-quinolinol-3-carboxylic acid (7-CQA) and 4-quinolinol-3-carboxylic acid (QA) were synthesized to investigate the antioxidant properties. 5,7-DCQA exhibited the highest ability to scavenge 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cationic radical (ABTS+.), 2,2'-diphenyl-1-picrylhydrazyl (DPPH) and galvinoxyl
Akio Kadowaki et al.
Chemistry and physics of lipids, 165(2), 178-185 (2012-01-03)
The antioxidative action of fullerenes has received much attention, but their reaction mechanism toward lipid-derived peroxyl radicals has not been well elucidated. In this study, the reaction products of [60]fullerene (C(60)) during the autoxidation of methyl linoleate (MeL) were isolated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service