Skip to Content
Merck
All Photos(1)

Key Documents

60983

Millipore

Kovac′s reagent for indoles

suitable for microbiology

Synonym(s):

4-(Dimethylamino)benzaldehyde solution, Indole reagent according to Kovac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Beilstein:
4132845
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

n-butanol
4-dimethylaminobenzaldehyde
hydrochloric acid

technique(s)

microbe id | specific enzyme detection: suitable

refractive index

n20/D 1.400-1.420

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

SMILES string

[H]C(=O)c1ccc(cc1)N(C)C

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kovac′s reagent for indoles is a solution of 4-(dimethylamino)benzaldehyde and hydrochloric acid in n-butanol.

Application

Kovac′s reagent for indoles has been used for enumerating E. coli O157:H7 in ground beef samples in different scenarios of stored, thawed, and as cooked patty samples. It has also been used for E coli enumeration in dewatered blackwater.

Biochem/physiol Actions

Kovac′s reagent detects the indole and aids in identifying the indole-positive and indole-negative microorganisms. E. coli can split tryptophan into indole and α-aminopropionic acid in the presence of the enzyme tryptophanase. p-Aminobenzaldehyde present in the reagent reacts with indole to form a cherry-red complex, that is soluble in alcohol, ether, and chloroform. It is recommended to use glucose free media and peptone with a high tryptophan content for the test.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ragnhildur Gunnarsdóttir et al.
Environmental science & technology, 46(22), 12408-12416 (2012-11-02)
Wastewater treatment in many Arctic regions is inadequate, even nonexisting. Natural freezing of wastewater in those areas may be beneficial for reduction of microorganisms. The aim of this study was to investigate the effect of long-term freezing, and repeated freezing
Stavros G Manios et al.
Meat science, 101, 25-32 (2014-12-03)
The effect of common handling practices (i.e., freezing, thawing and cooking) of beef patties on the survival of Salmonella spp. and Escherichia coli O157:H7, was evaluated. Inoculated ground beef was stored at -22 °C for 5 and 75 days. After
G L Lombard et al.
Journal of clinical microbiology, 18(3), 609-613 (1983-09-01)
Three reagents for detecting indole, Kovac, Ehrlich, and p-dimethylaminocinnamaldehyde (DMCA), were evaluated with commercial microtest systems for characterizing and identifying anaerobic bacteria. The DMCA reagent, the most sensitive of the three reagents, gave a positive reaction with 445 of 449
Rosario Zamora et al.
Journal of agricultural and food chemistry, 52(13), 4166-4171 (2004-06-24)
The Ehrlich reaction was optimized to determine the formation of pyrrolized phospholipids in edible oils in an attempt to understand the color reversion produced during the deodorization of poorly degummed edible oils. The procedure consisted of the treatment of the
M Stefek et al.
Biochimica et biophysica acta, 1502(3), 398-404 (2000-11-09)
In the present work, pepsin digests of tail tendons from streptozotocin-diabetic rats were found to contain material that reacted rapidly at room temperature with p-dimethylaminobenzaldehyde (Ehrlich's reagent) to give an adduct with an absorbance spectrum characteristic of the Ehrlich chromogen

Articles

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service