Skip to Content
Merck
All Photos(1)

Key Documents

60220-M

Sigma-Aldrich

Potassium phosphate monobasic

buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5-100.5%
form:
crystalline
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous
puriss. p.a.

Agency

reag. ISO
reag. Ph. Eur.

Assay

99.5-100.5%

form

crystalline

impurities

≤0.001% total nitrogen (N)

loss

≤0.2% loss on drying, 105 °C

pH

4.2-4.5 (20 °C, 5%)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

bp

>450 °C

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤15 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate monobasic is a soluble salt employed as heterogeneous catalyst, additive, fungicide, buffer, and plant fertilizer.

Application

Potassium phosphate monobasic can be used as a catalyst:
  • In the regioselective monobromination of aralkyl ketones to synthesize monobrominated ketones.
  • To synthesize acridine-1,8-diones and hexahydroquinolines via Hantzsch type reaction.
It can also be used as an additive in the oxidation of alcohols to aldehydes. The role of KH2PO4 was to regulate the pH of the solution, which affected the rate at which hydroxyl radicals were generated. As a result, the reaction′s selectivity was improved.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service