Skip to Content
Merck
All Photos(1)

Key Documents

60137

Supelco

Potassium chloride solution

~3 M KCl, saturated with silver chloride

Synonym(s):

Potassium chloride solution saturated with AgCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.61
grade:
for analytical purposes

grade

for analytical purposes

Quality Level

quality

saturated with silver chloride

concentration

~3 M KCl

density

1.13 g/mL at 20 °C

SMILES string

[K+].[Cl-]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

KCl exists as a white ionic solid, and can be prepared by neutralizing HCl with potassium hydroxide solution. It is less soluble in hot water as compared to cold water. It occurs naturally as the minerals carnallite and sylvine. It is generally used as a fertilizer.
Visit our Sensor Applications portal to learn more.

Application

Electrolyte filling solution for Ag/AgCl reference electrodes

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Daintith
Chemistry (Weinheim An Der Bergstrasse, Germany), 223-223 (2014)
Towards a fast, high specific and reliable discrimination of bacteria on strain level by means of SERS in a microfluidic device.
Walter A.
Lab on a chip, 11 (6), 1013-1021 (2011)
Masatoshi Takagaki et al.
Experimental neurology, 252, 12-17 (2013-11-20)
Sedatives in the neurointensive care unit can strongly influence patients' risks of developing secondary brain damage. In particular, isoflurane, a volatile anesthetic, has been recently re-introduced to the neurointensive care unit, and first clinical studies suggest beneficial effects due to
Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Yuanyuan Qu et al.
Nucleic acids research, 41(10), 5263-5272 (2013-04-13)
Bacterial nucleoid-associated proteins, such as H-NS-like proteins in Enterobacteriaceae, are abundant DNA-binding proteins that function in chromosomal DNA organization and gene transcription regulation. The Mycobacterium tuberculosis Lsr2 protein has been proposed to be the first identified H-NS analogue in Gram-positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service