58552
2-Methylpropene
purum, ≥99.5%
Synonym(s):
Isobutylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2C=CH2
CAS Number:
Molecular Weight:
56.11
Beilstein:
773645
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
2 (vs air)
vapor pressure
3278 mmHg ( 37.7 °C)
grade
purum
Assay
≥99.5%
autoignition temp.
869 °F
bp
−6.9 °C (lit.)
SMILES string
CC(C)=C
InChI
1S/C4H8/c1-4(2)3/h1H2,2-3H3
InChI key
VQTUBCCKSQIDNK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Cylinder with net ~3.6 kg
DIN 477 nr.1
Other Notes
Sales restrictions may apply
Recommended products
Z742161 DIN 1 Regulator OR 99112 DIN 1 Valve
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-112.0 °F - closed cup
Flash Point(C)
-80 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A theoretical investigation of the gas-phase oxidation reaction of the saturated tert-butyl radical.
Jong-Ho Choi et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 7(12), 2526-2532 (2006-11-09)
The radical-radical reaction mechanisms and dynamics of ground-state atomic oxygen [O(3P)] with the saturated tert-butyl radical (t-C4H9) are investigated using the density functional method and the complete basis set model. Two distinctive reaction pathways are predicted to be in competition:
Nava Dayan et al.
Journal of cosmetic science, 60(1), 15-24 (2009-03-20)
Hydrogenated polyisobutene (HP) is used in topically applied cosmetic/personal care formulations as an emollient that leaves a pleasing skin feel when applied, and rubbed in after application. This effect, although distinguishable to the user, is difficult to define and quantify.
Jiwen Li et al.
Se pu = Chinese journal of chromatography, 22(1), 81-83 (2005-02-17)
A capillary column HP-AlO2O3/S PLOT(50 m x 0.32 mm i.d. x 8 microm) has been used for the analysis of trace hydrocarbon impurities in high purity isobutene. The results showed that these hydrocarbon impurities could be well separated and with
Markus Burkhardt et al.
Langmuir : the ACS journal of surfaces and colloids, 24(5), 1769-1777 (2008-01-30)
We report on interpolyelectrolyte complexes (IPECs) formed by micelles of ionic amphiphilic diblock copolymers with polyisobutylene (PIB) and poly(sodium methacrylate) (PMANa) blocks interacting with quaternized poly(4-vinylpyridine) (P4VPQ). The interpolyelectrolyte complexation was followed by turbidimetry and small angle neutron scattering (SANS).
Nicolas Cottenye et al.
Chemical communications (Cambridge, England), 48(20), 2615-2617 (2012-02-02)
The polymer modification of short nucleotide sequences has been achieved for future use as self-assembled biologically active structures with sizes in the nanometre range. Co-assembly of the resulting DNA-based amphiphilic block copolymers with native proteins demonstrates the self-assembly of biological-like
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service