Skip to Content
Merck
All Photos(2)

Key Documents

49777

Supelco

Neotame

analytical standard

Synonym(s):

N-[N-(3,3-Dimethylbutyl)-L-α-aspartyl]-L-phenylalanine 1-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N2O5
CAS Number:
Molecular Weight:
378.46
Beilstein:
8352678
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E961
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -41.0±3.0°, c = 0.5 in H2O

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(O)=O)NCCC(C)(C)C

InChI

1S/C20H30N2O5/c1-20(2,3)10-11-21-15(13-17(23)24)18(25)22-16(19(26)27-4)12-14-8-6-5-7-9-14/h5-9,15-16,21H,10-13H2,1-4H3,(H,22,25)(H,23,24)/t15-,16-/m0/s1

InChI key

HLIAVLHNDJUHFG-HOTGVXAUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neotame is a white colored, dipeptide methyl ester, which is found to be a highly potent and non-nutritive sweetener or flavour enhancer and hence finds applications in variety of foods. It is manufactured from aspartame and 3,3-dimethylbutyraldehyde in a one-step chemical synthesis process.
Neotame is chemically related to aspartame, an artificial sweetener, used in the production of sugar-free low-calorie foodstuffs. It is approved as a food additive depending upon the food or beverage application.

Application

Neotame may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Wine samples using reversed-phase high-performance liquid chromatography (RP-HPLC).
  • Wastewaters using hydrophilic interaction liquid chromatography (HILIC).
  • Food samples using high-performance liquid chromatography (HPLC).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modifying the temporal profile of the high-potency sweetener neotame
Prakash I, et al.
Journal of Agricultural and Food Chemistry, 49, 786-789 (2001)
Neotame: discovery, properties, utility
Nofre C and Tinti M-J
Food Chemistry, 69, 245-257 (2000)
Neotame as a sweetener and flavour enhancer?Scientific Opinion of the Panel on Food Additives, Flavourings, Processing Aids and Materials in Contact with Food
European Food Safety Authority (EFSA)
EFSA Journal, 5(11), 581-581 (2007)
Determination of eight artificial sweeteners in wastewater by hydrophilic interaction liquid chromatography-tandem mass spectrometry
Kokotou GM and Thomaidis SN
Analytical Methods : Advancing Methods and Applications, 5(16), 3825-3833 (2013)
Determination of nine high-intensity sweeteners in various foods by high-performance liquid chromatography with mass spectrometric detection
Zygler A, et al.
Analytical and Bioanalytical Chemistry, 400(07), 2159-2172 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service