Skip to Content
Merck
All Photos(2)

Key Documents

49547

Supelco

Naphthol Yellow S

analytical standard

Synonym(s):

2,4-Dinitro-1-naphthol-7-sulfonic acid sodium salt, 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid sodium salt, Acid Yellow 1, Flavianic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4N2Na2O8S
CAS Number:
Molecular Weight:
358.19
Colour Index Number:
10316
Beilstein:
3839220
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[O-]c1c(cc([N+]([O-])=O)c2ccc(cc12)S([O-])(=O)=O)[N+]([O-])=O

InChI

1S/C10H6N2O8S.2Na/c13-10-7-3-5(21(18,19)20)1-2-6(7)8(11(14)15)4-9(10)12(16)17;;/h1-4,13H,(H,18,19,20);;/q;2*+1/p-2

InChI key

CTIQLGJVGNGFEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Naphthol Yellow S is a dye that is known for its hazardous nature.

Application

Naphthol Yellow S may be used as an analytical reference standard for the determination of Naphthol Yellow S in:
  • Foodstuffs by solid phase extraction (SPE) and high performance liquid chromatography (HPLC)-diode array detection (DAD)-ion trap time-of-flight tandem mass spectrometry (IT-TOF-MS3) equipped with multiple reaction monitoring (MRM) detection.
  • Drinks and candies by SPE followed by analysis using HPLC coupled to photodiode array (PDA) detector.
  • Wastewater samples by adsorption column chromatography.
  • Embroideries designed by the famous 19th century French painter Emile Bernard.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and determination of 34 water-soluble synthetic dyes in foodstuff by high performance liquid chromatography-diode array detection-ion trap time-of-flight tandem mass spectrometry.
Li XQ, et al.
Food Chemistry, 182(9), 316-326 (2015)
High-performance liquid chromatography and non-destructive three-dimensional fluorescence analysis of early synthetic dyes.
van Bommel MR, et al.
Journal of Chromatography A, 1157(1-2), 260-272 (2007)
Photodegradation of hazardous dye naphthol yellow s over titanium dioxide.
Jain R and Sikarwar S
Journal of Dispersion Science and Technology, 32(9), 1345-1352 (2011)
W M Frederiks et al.
Histochemistry, 68(1), 49-53 (1980-01-01)
The purpose of this study is to compare the protein content of parenchymal and non-parenchymal nuclei, as isolated from rat liver. The nuclei have been separated by means of a 1 g-sedimentation technique. The protein content of the separated nuclei
J Tas et al.
Acta histochemica. Supplementband, 20, 69-73 (1979-01-01)
After staining with Naphthol Yellow S (NYS) at optimal conditions of pH (2.8), the protein content of rat liver cells isolated by means of a collagenase perfusion technique was found to be cytophotometrically immeasurable, because of too high local dye

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service