The suggested approach to ascertain its impact is to conduct a controlled side-by-side experiment to determine if its presence has any adverse effects.
48917-U
ProClin™ 300
pkg of 3.6 L, ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 3 %
Synonym(s):
3% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one
About This Item
Recommended Products
Quality Level
form
liquid
shelf life
3 yr
contains
Alkyl Carboxylate
composition
CMIT/MIT, 3%
packaging
pkg of 3.6 L
parameter
<45 °C operating temperature
color
colorless to light yellow
matrix
Modified glycol
operating pH range
2.5-8.5
pH
4.1 (100 g/L)
bp
189 °C
mp
-40 °C
solubility
water: soluble
density
1.03 g/cm3
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
244.4 °F - closed cup
Flash Point(C)
118 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What are the effects of Proclin 300 on PCR?
1 answer-
Helpful?
-
-
Is there specific gravity data available for Proclin 300 at 25°C, considering it's listed as 1.03 g/ml at 22°C?
1 answer-
There is no available information regarding the density of ProClin 300 at 25°C. The value of 1.03 g/ml at 22°C is accurate.
Helpful?
-
-
What is the matrix of the ProClin 300?
1 answer-
ProClin 300 is a viscous solution containing 93-95 % Modified glycol and 2-3 % Alkyl carboxylate.
Helpful?
-
-
What are the active ingredients in ProClin 300?
1 answer-
ProClin 300 contains two active ingredients: 5-Chloro-2-methyl-4-isothiazolin-3-one (CMIT, CAS RN 26172-55-4) and 2-Methyl-4-isothiazolin-3-one (MIT, CAS RN 2682-20-4).
Helpful?
-
-
What is the density of ProClin 300?
1 answer-
The density of ProClin 300 is reported as 8.59 pounds per gallon. The specific gravity at 22°C is reported as 1.03 g/mL.
Helpful?
-
-
What is the concentration of active ingredients in ProClin 300?
1 answer-
ProClin 300 contains a total of 3% active ingredient.
Helpful?
-
-
When would I choose to use ProClin 300?
1 answer-
ProClin 300 is best used with production runs of standards, controls, and large volume buffers and solutions. The product is quite viscous and difficult to work with when trying to measure small volumes.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service