Skip to Content
Merck
All Photos(2)

Key Documents

46431

Supelco

Boldenone

VETRANAL®, analytical standard

Synonym(s):

1-Dehydrotestosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Beilstein:
6549081
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h7,9,11,14-17,21H,3-6,8,10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1

InChI key

RSIHSRDYCUFFLA-DYKIIFRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Piper et al.
Drug testing and analysis, 2(5), 217-224 (2010-05-15)
Boldenone (androsta-1,4-dien-17beta-ol-3-one, Bo) is an anabolic steroid known to have been used in cattle breeding or equine sport as a doping agent for many years. Although not clinically approved for human application, Bo or its main metabolite 5beta-androst-1-en-17beta-ol-3-one (BM1) were
Silvana Casati et al.
Steroids, 74(2), 250-255 (2008-12-17)
Boldenone is an androgenic anabolic steroid intensively used for growth promoting purposes in animals destined for meat production and as a performance enhancer in athletics. Therefore its use is officially banned either in animals intended for consumption or in humans.
C Gómez et al.
The Journal of steroid biochemistry and molecular biology, 132(3-5), 239-246 (2012-06-06)
Boldenone is one of the most frequently detected anabolic androgenic steroids in doping control analysis. Boldenone misuse is commonly detected by the identification of the active drug and its main metabolite, 5β-androst-1-en-17β-ol-3-one (BM1), by gas chromatography-mass spectrometry (GC-MS), after previous
I Thiblin et al.
Forensic science international, 184(1-3), e7-11 (2008-12-27)
The use of anabolic androgenic steroids (AAS) has been associated with different adverse effects, some of which potentially lethal. Most users of AAS are male, but the prevalence of such use appears to be increasing in females. Here we present
Philip B Grace et al.
Rapid communications in mass spectrometry : RCM, 22(19), 2999-3007 (2008-09-09)
Following administration of the anabolic steroid 19-nortestosterone or its esters to the horse, a major urinary metabolite is 19-nortestosterone-17beta-sulphate. The detection of 19-nortestosterone in urine from untreated animals has led to it being considered a naturally occurring steroid in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service