Skip to Content
Merck
All Photos(1)

Documents

46267

Supelco

Trichloroethylene

analytical standard, stabilized with 30 – 50 ppm Diisopropylamine

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

autoignition temp.

770 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroethylene is a volatile and chlorinated organic compound, which finds applications as a solvent and greasing agent.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product availableon our online platform ChemisTwin® for NMR. You can use thisdigital equivalent on ChemisTwin® for your sample identityconfirmation and compound quantification (with digital external standard). AnNMR spectrum of this substance can be viewed and an online comparison againstyour sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenol and trichloroethylene degradation by Pseudomonas cepacia G4: kinetics and interactions between substrates.
Folsom R B, et al.
Applied and Environmental Microbiology, 56(5), 1279-1285 (1990)
Michihiro Kamijima et al.
International archives of occupational and environmental health, 80(5), 357-370 (2006-11-16)
Workers exposed to trichloroethylene (TCE) rarely show severe generalized skin disorders and accompanying hepatitis which resemble drug hypersensitivities. The disorders are completely different from solvent-induced irritating contact dermatitis, and their serious consequences have become one of the critical occupational health
Cheryl Siegel Scott et al.
Environmental health perspectives, 114(9), 1471-1478 (2006-09-13)
A large body of epidemiologic evidence exists for exploring causal associations between cancer and trichloroethylene (TCE) exposure. The U.S. Environmental Protection Agency 2001 draft TCE health risk assessment concluded that epidemiologic studies, on the whole, support associations between TCE exposure
Luoping Zhang et al.
Carcinogenesis, 34(4), 799-802 (2013-01-02)
Trichloroethylene (TCE) has been associated with a variety of immunotoxic effects and may be associated with an increased risk of non-Hodgkin lymphoma (NHL). Altered serum immunoglobulin (Ig) levels have been reported in NHL patients and in animals exposed to TCE.
Jane C Caldwell et al.
Environmental health perspectives, 114(9), 1457-1463 (2006-09-13)
Trichloroethylene (TCE) exposure has been associated with increased risk of liver and kidney cancer in both laboratory animal and epidemiologic studies. The U.S. Environmental Protection Agency 2001 draft TCE risk assessment concluded that it is difficult to determine which TCE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service