Skip to Content
Merck
All Photos(1)

Key Documents

46208

Supelco

(+)-Fenchone

analytical standard

Synonym(s):

(+)-1,3,3-Trimethyl-2-norbornanone, (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2206555
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (sum of enantiomers, GC)

optical activity

[α]20/D +62±1°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.462

bp

63-65 °C/13 mmHg (lit.)

mp

5-7 °C (lit.)

density

0.945 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1(C)[C@@H]2CC[C@@](C)(C2)C1=O

InChI

1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

LHXDLQBQYFFVNW-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Highly purified (+)-fenchone; the keto function can be transformed in many ways

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

P. Gosselin et al.
Tetrahedron Letters, 31, 3151-3151 (1990)
L.A. Paquette et al.
The Journal of Organic Chemistry, 57, 3965-3965 (1992)
Olga Tzakou et al.
Natural product communications, 4(8), 1103-1106 (2009-09-23)
The essential oils from leaves and inflorescences of L. cariensis Boiss. and L. stoechas L. subsp. stoechas collected in Greece were analyzed by GC and GC/MS. In the inflorescences and leaves essential oils of L. cariensis the most abundant metabolite
B Simándi et al.
Journal of agricultural and food chemistry, 47(4), 1635-1640 (1999-11-24)
Ground fennel seeds were extracted with supercritical carbon dioxide. Small-scale subsequent extractions of the same sample showed that the composition of volatile compounds was changed with the extension of extraction time and only principal volatile components (limonene, fenchone, methylchavicol, and
I M Isa et al.
Talanta, 87, 230-234 (2011-11-22)
A new cobalt(II) ion selective electrode based on palladium(II) dichloro acetylthiophene fenchone azine(I) has been developed. The best membrane composition is found to be 10:60:10:21.1 (I)/PVC/NaTPB/DOP (w/w). The electrode exhibits a Nerstian response in the range of 1.0 × 10(-1)-1.0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service