Skip to Content
Merck
All Photos(1)

Documents

45776

Supelco

Dibenzothiophene

analytical standard

Synonym(s):

DBT, Diphenylene sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8S
CAS Number:
Molecular Weight:
184.26
Beilstein:
121101
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

332-333 °C (lit.)

mp

97-100 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc2c(c1)sc3ccccc23

InChI

1S/C12H8S/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

IYYZUPMFVPLQIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adeniyi S Ogunlaja et al.
Dalton transactions (Cambridge, England : 2003), 41(45), 13908-13918 (2012-10-02)
The reaction between [V(IV)OSO(4)] and the tetradentate N(2)O(2)-donor Schiff base ligand, N,N-bis(o-hydroxybenzaldehyde)phenylenediamine (sal-HBPD), obtained by the condensation of salicylaldehyde and o-phenylenediamine in a molar ratio of 2 : 1 respectively, resulted in the formation of [V(IV)O(sal-HBPD)]. The molecular structure of
Ellen M Cooper et al.
Environmental toxicology and chemistry, 29(11), 2409-2416 (2010-09-24)
Biodegradation of pollutants often results in incomplete mineralization and formation of degradation products with unknown chemical and toxicological characteristics. Ultraviolet (UV) irradiation, a common technology used in water and wastewater treatment, may help reduce aqueous concentrations of degradation products produced
Shilpi Aggarwal et al.
Molecular bioSystems, 8(10), 2724-2732 (2012-07-27)
Rhodococcus erythropolis has been widely studied for desulfurization. However, activity levels required for commercial application have not been achieved. A major limitation of the current work in biodesulfurization is inadequate information regarding sulfur metabolism generally, and in particular the metabolism
Palladium-catalyzed double C-H activation directed by sulfoxides in the synthesis of dibenzothiophenes.
Rajarshi Samanta et al.
Angewandte Chemie (International ed. in English), 50(22), 5217-5220 (2011-04-21)
Christian Zeigler et al.
Analytical chemistry, 84(5), 2245-2252 (2012-02-22)
Polycyclic aromatic sulfur heterocycles (PASH) are sulfur analogues of polycyclic aromatic hydrocarbons (PAH). Alkylated PAH attract much attention as carcinogens, mutagens, and as diagnostics for environmental forensics. PASH, in contrast, are mostly ignored in the same studies due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service