Skip to Content
Merck
All Photos(1)

Documents

45705

Supelco

Vinclozolin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9Cl2NO3
CAS Number:
Molecular Weight:
286.11
Beilstein:
8331312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC1(OC(=O)N(C1=O)c2cc(Cl)cc(Cl)c2)C=C

InChI

1S/C12H9Cl2NO3/c1-3-12(2)10(16)15(11(17)18-12)9-5-7(13)4-8(14)6-9/h3-6H,1H2,2H3

InChI key

FSCWZHGZWWDELK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seongeon Lee et al.
Marine pollution bulletin, 113(1-2), 253-257 (2016-10-04)
Since the ban on tributyltin (TBT) in 2008, dozens of alternative antifouling agents have been used instead. Unfortunately, these alternative antifouling agents contain diverse toxic components, which have contaminated the coasts. From 2006 to 2013, the concentration of chlorothalonil, dichlofluanid
Teresa L Dzieweczynski et al.
Environmental pollution (Barking, Essex : 1987), 235, 1015-1021 (2018-01-26)
Components of boldness, such as activity level and locomotion, influence an individual's ability to avoid predators and acquire resources, generating fitness consequences. The presence of endocrine disrupting chemicals (EDCs) in the aquatic environment may affect fitness by changing morphology or
Abdelali Lehraiki et al.
Reproductive toxicology (Elmsford, N.Y.), 31(4), 424-430 (2010-12-22)
Continuous, low-dose exposure to a phytoestrogen (1 mg/kg/day genistein) and/or to an antiandrogenic food contaminant (1 mg/kg/day vinclozolin) has been recently reported to affect male reproductive tract and fertility [1] in adults. We investigated whether alterations of the testis are
Alison M Anderson et al.
PloS one, 7(4), e34158-e34158 (2012-04-13)
Endocrine disruptor chemicals elicit adverse health effects by perturbing nuclear receptor signalling systems. It has been speculated that these compounds may also perturb epigenetic mechanisms and thus contribute to the early origin of adult onset disease. We hypothesised that histone
Justin M Conley et al.
Toxicological sciences : an official journal of the Society of Toxicology, 164(1), 166-178 (2018-06-27)
Biomonitoring efforts have clearly shown that all humans are exposed to chemical mixtures. Of concern is whether or not exposure to mixtures during pregnancy contributes to congenital abnormalities in children even when each chemical is at an individual dose that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service