Skip to Content
Merck
All Photos(1)

Key Documents

45639

Supelco

Propanil

PESTANAL®, analytical standard

Synonym(s):

3′,4′-Dichloropropionanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Cl2NO
CAS Number:
Molecular Weight:
218.08
Beilstein:
2365645
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(=O)Nc1ccc(Cl)c(Cl)c1

InChI

1S/C9H9Cl2NO/c1-2-9(13)12-6-3-4-7(10)8(11)5-6/h3-5H,2H2,1H3,(H,12,13)

InChI key

LFULEKSKNZEWOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propanil is a contact, post-emergence herbicide, used in the cultivation of irrigated rice. Its mode of action involves controlling, the electron transport inhibition of photosynthesis in herbs of wide leaves.

Application

Propanil may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen B Pruett et al.
Toxicological sciences : an official journal of the Society of Toxicology, 109(2), 265-275 (2009-04-10)
Previous studies have demonstrated that the stress response induced by some drugs and chemicals contributes in a predictable way to alteration of particular immunological parameters in mice. It has not been determined if mice can become tolerant or habituated with
Gary O Rankin et al.
Environmental toxicology, 23(4), 435-442 (2008-01-25)
Propanil is a postemergence herbicide used primarily in rice and wheat production in the United States. The reported toxicities for propanil exposure include methemoglobinemia, immunotoxicity, and nephrotoxicity. A major metabolite of propanil, 3,4-dichloroaniline (3,4-DCA), has been shown to be a
Joana Damásio et al.
Ecotoxicology (London, England), 19(6), 1084-1094 (2010-04-07)
The aim of this investigation was to identify major pesticides that may cause detrimental effects in bivalve species affected by agricultural pollution. Investigations were carried out using freshwater clams (Corbicula fluminea) transplanted in the main drainage channels that collect the
Jun Zhang et al.
International journal of systematic and evolutionary microbiology, 62(Pt 3), 495-499 (2011-04-12)
A novel facultatively anaerobic, non-spore-forming, non-motile, catalase- and oxidase-positive, Gram-negative and rod-shaped bacterial strain, designated Y12(T), was isolated from activated sludge of a wastewater bio-treatment facility. The strain was able to degrade about 90% of added propanil (100 mg l(-1))
Joana Luísa Pereira et al.
Ecotoxicology and environmental safety, 68(3), 386-396 (2006-12-08)
The widespread increase of pesticides application in crops frequently leads to the contamination of vicinal freshwater lentic ecosystems. Herbicides such as propanil may impair cladoceran fitness, which is per se strongly influenced by the food availability and/or its acquisition efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service