Skip to Content
Merck
All Photos(2)

Key Documents

45467

Supelco

Edifenphos

PESTANAL®, analytical standard

Synonym(s):

O-Ethyl S,S-diphenyl phosphorodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15O2PS2
CAS Number:
Molecular Weight:
310.37
Beilstein:
1988797
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(Sc1ccccc1)Sc2ccccc2

InChI

1S/C14H15O2PS2/c1-2-16-17(15,18-13-9-5-3-6-10-13)19-14-11-7-4-8-12-14/h3-12H,2H2,1H3

InChI key

AWZOLILCOUMRDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Edifenphos is a selective and protective, organophosphorus foliar fungicide, used in controlling rice diseases such as blast caused by Pyricularia oryzae, sheath blight caused by Rhizoctonia solani and brown spot by Cochliobolus miyabeanus.

Application

Edifenphos may be used as a reference standard in the analysis of the analyte in rice using gold nanoparticle (AuNP)-based colorimetric multi-aptasensor.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry of Cell Walls and Membranes in Fungi (2012)
Edifenphos, Inhibitor of Phosphatidylcholine Biosynthesis in Pyricularia oryzae.
Kodama O, et al.
Agricultural and Biological Chemistry, 44(5), 1015-1021 (1980)
Detection of Iprobenfos and Edifenphos using a new Multi-aptasensor.
Agricultural and Biological Chemistry, 868(5), 60-66 (2015)
The Pesticide Encyclopedia (2014)
J R Math et al.
Indian journal of experimental biology, 36(1), 39-42 (1998-04-16)
Exposure of rats to 0.1, 0.15 and 0.20 mg/100 g body weight edifenphos (i.p.) had no significant effect on the number of healthy follicles and atretic follicles in all the stages. However, treatment with 0.25 mg dose resulted in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service