Skip to Content
Merck
All Photos(1)

Key Documents

45377

Supelco

Chlorobromouron

PESTANAL®, analytical standard

Synonym(s):

Chlorbromuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BrClN2O2
CAS Number:
Molecular Weight:
293.54
Beilstein:
2128736
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CON(C)C(=O)Nc1ccc(Br)c(Cl)c1

InChI

1S/C9H10BrClN2O2/c1-13(15-2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)

InChI key

NLYNUTMZTCLNOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hygienic standardization of the herbicide Maloran in the air of a work area].
G A Voĭtenko et al.
Gigiena i sanitariia, (8)(8), 76-78 (1981-08-01)
J Clemy Monicka et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 94, 30-35 (2012-04-21)
FT-Raman and IR spectra of the herbicidal compound chlorbromuron have been recorded and analyzed. The detailed interpretation of the vibrational spectra has been carried out with the aid of normal coordinate analysis (NCA) following the scaled quantum mechanical force field
Susana Silva Martínez et al.
Water research, 43(1), 33-40 (2008-11-07)
The removal of low concentration of chlorbromuron herbicide in aqueous systems was carried out by electro-Fenton process comprised of three-electrode divided and undivided cell with a reticulated vitreous carbon cathode and platinum anode. The electro-Fenton was also carried out in
[Comparative gonadotoxic effect of maloran in laboratory animals after its administration by different routes].
N R Shepel'skaia et al.
Gigiena i sanitariia, (3)(3), 84-85 (1985-03-01)
Liying Guo et al.
Mikrochimica acta, 186(7), 464-464 (2019-06-24)
Graphene oxide was covalently modified with p-phenylenediamine via a diazonium reaction. The resulting material was employed as a sorbent for the solid-phase extraction of six phenylurea herbicides (metoxuron, monuron, chlortoluron, isoproturon, monolinuron, and buturon) from environmental water and lettuce leafs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service