Skip to Content
Merck
All Photos(1)

Key Documents

45331

Supelco

Azamethiphos

PESTANAL®, analytical standard

Synonym(s):

S-((6-Chloro-2-oxooxazolo[4,5-b]pyridin-3(2H)-yl)methyl) O,O-dimethyl phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10ClN2O5PS
CAS Number:
Molecular Weight:
324.68
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(OC)SCN1C(=O)Oc2cc(Cl)cnc12

InChI

1S/C9H10ClN2O5PS/c1-15-18(14,16-2)19-5-12-8-7(17-9(12)13)3-6(10)4-11-8/h3-4H,5H2,1-2H3

InChI key

VNKBTWQZTQIWDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Azamethiphos is classified under the heterocyclic organothiophosphate group of compounds.

Application

Azamethiphos may be used as an analytical reference standard for the determination of azamethiphos in:
  • Salmon feed samples by ultra-high performance liquid chromatography combined with traveling-wave ion mobility/quadrupole time-of-flight mass spectrometry (UHPLC-TWIMS-QTOFMS).
  • Surface water samples by solid-phase extraction (SPE) and LC coupled to quadrupole-Orbitrap high resolution tandem mass spectrometry (Q-Orbitrap-MS).
  • Fish fillet samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and LC-QTOFMS.
  • Human serum samples by QuEChERS combined with LC coupled to tandem mass spectrometry (MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anders Fallang et al.
Pest management science, 60(12), 1163-1170 (2004-12-04)
Acetylcholinesterase (AChE) is the target of a major pesticide family, the organophosphates, which were extensively used as control agents of sea lice on farmed salmonids in the early 1990s. From the mid-1990s the organophosphates dichlorvos and azamethiphos were seriously compromised
W Ernst et al.
Marine pollution bulletin, 42(6), 433-444 (2001-07-27)
Pesticides are used extensively in the finfish aquaculture industry to control sea lice infestations on farmed salmon. The most prevalent method of use is to enclose a net pen with an impervious tarpaulin and mix a pesticide solution within that
L E Burridge et al.
Ecotoxicology and environmental safety, 43(2), 165-169 (1999-06-22)
The pesticide formulation Salmosan (47.5% w/w azamethiphos) is currently registered for use, in Canada, to treat salmonids for infestations of the copepod parasites, Lepeophtheirus salmonis and Caligus elongatus (sea lice). Determination was made of the acute lethality of this product
Targeted approach for qualitative screening of pesticides in salmon feed by liquid chromatography coupled to traveling-wave ion mobility/quadrupole time-of-flight mass spectrometry.
Regueiro J, et al.
Food Control, 78(4), 116-125 (2017)
P J Collins et al.
Journal of economic entomology, 93(4), 1357-1363 (2000-09-14)
Four organophosphate insecticides, azamethiphos, fenitrothion, chlorpyrifos-methyl, and pirimiphos-methyl, were tested as surface treatments on concrete (porous surface) and galvanized steel (nonporous surface) panels (0.3 by 0.3 m) against adults of three Liposcelid psocid spp.--Liposcelis bostrychophila Badonnel, Liposcelis entomophila (Enderlein), and

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service