Skip to Content
Merck
All Photos(2)

Key Documents

43073

Supelco

Paracetamol β-D-glucuronide

analytical standard

Synonym(s):

p-Acetamidophenyl β-D-glucuronide, Acetaminophen glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO8
CAS Number:
Molecular Weight:
327.29
Beilstein:
46644
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(NC(C)=O)C=C2)O[C@H](C(O)=O)[C@H]1O

InChI

1S/C14H17NO8/c1-6(16)15-7-2-4-8(5-3-7)22-14-11(19)9(17)10(18)12(23-14)13(20)21/h2-5,9-12,14,17-19H,1H3,(H,15,16)(H,20,21)/t9-,10-,11+,12-,14+/m0/s1

InChI key

IPROLSVTVHAQLE-BYNIDDHOSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sifeng Mao et al.
Lab on a chip, 12(1), 219-226 (2011-11-19)
In this work, we developed a microfluidic device for the imitation of drug metabolism in human liver and its cytotoxicity on cells. The integrated microfluidic device consists of three sections: (1) bioreactors containing poly(ethylene) glycol (PEG) hydrogel encapsulated human liver
Nicole J Barshop et al.
Journal of pediatric gastroenterology and nutrition, 52(2), 198-202 (2011-01-18)
: The aim of the study was to evaluate UDP-glucuronyltransferase activity and the pharmacokinetics of a single oral dose of acetaminophen (APAP) in children with nonalcoholic fatty liver disease (NAFLD). : Twelve boys 10 to 17 years old with biopsy-proven
Karel Allegaert et al.
Therapeutic drug monitoring, 31(4), 411-415 (2009-06-06)
Compared with phase I isoenzymes, data on isoenzyme-specific phenotypic activity of uridine diphosphate glucuronosyltransferase (UGT) and its covariates in neonates are limited. In vivo observations on morphine, paracetamol (acetaminophen), and propofol disposition throughout childhood confirm the overall low-glucuronidation activity in
Carolina I Ghanem et al.
Biochemical pharmacology, 77(10), 1621-1628 (2009-05-12)
Development of resistance to toxic effects of acetaminophen (APAP) was reported in rodents and humans, though the mechanism is only partially understood. We examined in rats the effect of administration with subtoxic daily doses (0.2, 0.3, and 0.6g/kg, i.p.) of
Jinchun Sun et al.
Drug metabolism letters, 3(3), 130-136 (2009-08-26)
A LC/MS-based metabolomic assay was utilized to investigate a drug's excretion kinetic profile in urine so that the drug toxicity information could be obtained. Groups of 10 male Sprague-Dawley rats per dose were orally gavaged with a single dose of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service