Skip to Content
Merck
All Photos(1)

Key Documents

41325

Sigma-Aldrich

Mayer′s Mucicarmine Stain Solution

for microscopy

Synonym(s):

Mucicarmine Stain Solution according to Mayer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
MA.02

grade

for microscopy

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

suitability

suitable for microscopy
fungi

application(s)

food and beverages
hematology
histology

storage temp.

room temp

General description

Most mucins contain serveral glycoproteins and can therefore be detected by stains for carbohydrates. Best carmine stain is specifically used for glycogens. Mucicarmine is an empirical stain. When applied for fungal staining, it stains the spores and hyphae as pale to dark red.

Components

Composition:
Carmine 1 g, Aluminum chloride 0.5 g, water 2 ml

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Flávia Gazzoni et al.
Mycopathologia, 167(4), 197-202 (2008-12-05)
Here we report an unusual case of disseminated cryptococcosis in a patient with AIDS. Although typical Cryptococcus neoformans micromorphology was observed in tongue biopsy, cervical lymph node examination revealed atypical histopathologic findings. These included pseudohyphae, chains of budding yeasts and
Oluwole Fadare
Pathology international, 56(11), 683-687 (2006-10-17)
Some examples of lobular carcinoma in situ (LCIS) may be composed in part of signet ring cells. Such proliferations have been considered examples of pleomorphic LCIS based on pathological features of the more conventional component. However, the occurrence of LCIS
O E Battles et al.
American journal of clinical pathology, 108(1), 6-12 (1997-07-01)
To identify a sensitive marker for extramammary Paget's disease and to identify histochemical and immunohistochemical features that suggest occult pelvic cancer in patients with extramammary Paget's disease, we retrieved all cases between 1983 and 1992 with a Standardized Nomenclature of
Ji Kon Ryu et al.
Diagnostic cytopathology, 31(2), 100-105 (2004-07-30)
Pancreatic cystic neoplasms comprise a pathologically heterogeneous group with many shared clinical features. We assessed the reliability of cyst fluid analysis for the differential diagnosis of pancreatic cysts. Cyst fluid was obtained by fine-needle aspiration from 78 pancreatic cysts. The
Abderrahman Ouban et al.
Clinical colorectal cancer, 1(4), 243-248 (2002-11-27)
Multidirectional differentiation in colorectal carcinomas is a rare phenomenon. Four cases are reported herein, and their clinical and pathologic characteristics are discussed. Two men and 2 women between the ages of 56 and 76 years who presented with abdominal symptoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service