Skip to Content
Merck
All Photos(1)

Documents

394874

Supelco

1-(Trimethylsilyl)imidazole

for GC derivatization, LiChropur

Synonym(s):

TSIM, N-Trimethylsilylimidazole, TMSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
Beilstein:
606148
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥94.0%

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.475 (lit.)

bp

93-94 °C/14 mmHg (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)n1ccnc1

InChI

1S/C6H12N2Si/c1-9(2,3)8-5-4-7-6-8/h4-6H,1-3H3

InChI key

YKFRUJSEPGHZFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Trimethylsilyl)imidazole (TMSIM) has high silyl donor ability. It does not react with amino groups and also does not help in formation of enol-ether on unprotected ketone groups. It is useful as a silylating agent for ecdysones, norepinephrine, dopamine, steroids, sugars, sugar phosphates and ketose isomers.
Powerful silylating agent, particularly for alcohols; Synthesis of acyl imidazolides

Application

1-(Trimethylsilyl) imidazole may be used for the synthesis an O-silylated derivative of a hexahomotriazacalix[3]arene. It may also be used for the silylation of Ti-MCM-41 catalyst, to study its effect on the olefin epoxidation with aqueous H2O2, 1-(Trimethylsilyl) imidazole was found to improve the hydrophobicity and the catalytic performance of Ti-MCM-41.
Learn more in the Product Information

Other Notes

Reagent for trimethylsilyl and trimethylsilyl ether.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silylation of Ti-MCM-41 by trimethylsilyl-imidazole and its effect on the olefin epoxidation with aqueous H2O2.
Jie Bu and Rhee HK.
Catalysis Letters, 66 (4), 245-249 (2000)
Synthesis and crystal structure of an O-silylated hexahomotriazacalix [3] arene.
Panadda C
Tetrahedron Letters, 39 (31), 5473-5476 (1998)
Letter: Analysis of bile acids by mass fragmentography -application of 1:1 mixture technique by use of stable isotope labeled compounds.
M Inoue et al.
Chemical & pharmaceutical bulletin, 22(8), 1949-1950 (1974-08-01)
Thomas J. Bruno, Paris D.N. Svoronos,
CRC Handbook of Basic Tables for Chemical Analysis, 111-111 (2010)
J.D. Nicholson
Analyst, 103, 193-193 (1978)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service